Merck
  • Doxorubicin-loaded polysaccharide nanoparticles suppress the growth of murine colorectal carcinoma and inhibit the metastasis of murine mammary carcinoma in rodent models.
Doxorubicin-loaded polysaccharide nanoparticles suppress the growth of murine colorectal carcinoma and inhibit the metastasis of murine mammary carcinoma in rodent models.
Biomaterials (2015-03-17)
Mingqiang Li, Zhaohui Tang, Dawei Zhang, Hai Sun, Huaiyu Liu, Ying Zhang, Yuanyuan Zhang, Xuesi Chen
ABSTRACT
As a synergistic drug combination, doxorubicin-loaded cisplatin crosslinked polysaccharide-based nanoparticles (Dex-SA-DOX-CDDP) have demonstrated enhanced antitumor efficacy and reduced systemic toxicity via optimized biodistribution, controlled drug release, prolonged blood circulation, and improved tolerability, compared to the non-crosslinked nanoparticles or free doxorubicin. Herein, we apply the Dex-SA-DOX-CDDP nanoparticles as an efficient antitumor agent to treat colorectal and breast tumors in three different in vivo models, i.e. subcutaneously implanted colorectal carcinoma, dimethylhydrazine-induced autochthonous colorectal carcinoma, and metastatic mammary carcinoma, which more closely simulate the natural milieu of the original tumor with intact pathological and immunological responses. Based on the properties of this combination in higher tumor accumulation and penetrating efficiency, the Dex-SA-DOX-CDDP nanoparticles significantly decreased the tumor sizes in CT26 cell line xenograft tumors compared to control. In addition, the affected animals' lifespan was significantly extended after the Dex-SA-DOX-CDDP treatment, in the autochthonous colon cancer model. Moreover, with the aid of iRGD, Dex-SA-DOX-CDDP could effectively block primary tumor growth and prevent the metastasis of 4T1 murine mammary carcinoma. In conclusion, Dex-SA-DOX-CDDP nanoparticles remarkably inhibit growth of colorectal carcinoma and metastasis of mammary carcinoma in vivo, which provides potential application as a safe and efficient antitumor agent in treatment of these cancers.
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt dihydrate, ≥97.0% (KT)
Sigma-Aldrich
4′,6-Diamidino-2-phenylindole dihydrochloride, suitable for fluorescence, BioReagent, ≥95.0% (HPLC)
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt dihydrate, meets analytical specification of Ph. Eur., BP, USP, FCC, 99.0-101.0%
Sigma-Aldrich
N,N-Dimethylhydrazine, 98%
Sigma-Aldrich
5,5-Dimethylhydantoin, 97%
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt dihydrate, BioUltra, ≥99.0% (T)
Sigma-Aldrich
4′,6-Diamidino-2-phenylindole dihydrochloride, powder, BioReagent, suitable for cell culture, ≥98% (HPLC), suitable for fluorescence
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt dihydrate, Sigma Grade, suitable for plant cell culture, 98.5-101.5%
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt dihydrate, reagent grade, 98.5-101.5% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt dihydrate, suitable for electrophoresis, Molecular Biology, 99.0-101.0% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt dihydrate, ACS reagent, 99.0-101.0%
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt dihydrate, BioUltra, 98.5-101.5%
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, 2%, solution
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, Molecular Biology, 0.5 M in H2O, DNase, RNase, NICKase and protease, none detected