Merck
  • Aging disrupts cell subpopulation dynamics and diminishes the function of mesenchymal stem cells.
Aging disrupts cell subpopulation dynamics and diminishes the function of mesenchymal stem cells.
Scientific reports (2014-11-22)
Dominik Duscher, Robert C Rennert, Michael Januszyk, Ersilia Anghel, Zeshaan N Maan, Alexander J Whittam, Marcelina G Perez, Revanth Kosaraju, Michael S Hu, Graham G Walmsley, David Atashroo, Sacha Khong, Atul J Butte, Geoffrey C Gurtner
ABSTRACT
Advanced age is associated with an increased risk of vascular morbidity, attributable in part to impairments in new blood vessel formation. Mesenchymal stem cells (MSCs) have previously been shown to play an important role in neovascularization and deficiencies in these cells have been described in aged patients. Here we utilize single cell transcriptional analysis to determine the effect of aging on MSC population dynamics. We identify an age-related depletion of a subpopulation of MSCs characterized by a pro-vascular transcriptional profile. Supporting this finding, we demonstrate that aged MSCs are also significantly compromised in their ability to support vascular network formation in vitro and in vivo. Finally, aged MSCs are unable to rescue age-associated impairments in cutaneous wound healing. Taken together, these data suggest that age-related changes in MSC population dynamics result in impaired therapeutic potential of aged progenitor cells. These findings have critical implications for therapeutic cell source decisions (autologous versus allogeneic) and indicate the necessity of strategies to improve functionality of aged MSCs.
Sigma-Aldrich
Isopropyl alcohol, ≥99.7%, FCC, FG
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, 98%
Sigma-Aldrich
2-Propanol, Laboratory Reagent, ≥99.5%
Sigma-Aldrich
Isopropyl alcohol, meets USP testing specifications
Sigma-Aldrich
2-Propanol, ACS reagent, ≥99.5%
Sigma-Aldrich
2-Propanol, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%
Sigma-Aldrich
2-Propanol, suitable for HPLC, 99.9%
Supelco
2-Propanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
2-Propanol, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
2-Propanol, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%, poly coated bottles
Sigma-Aldrich
2-Propanol, BioReagent, ≥99.5%, Molecular Biology
Sigma-Aldrich
2-Propanol, anhydrous, 99.5%
Supelco
2-Propanol, analytical standard
Sigma-Aldrich
2-Propanol, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
2-Propanol, puriss. p.a., ACS reagent, ≥99.8% (GC)
Sigma-Aldrich
2-Propanol, suitable for HPLC, 99.5%
USP
2-Propanol, United States Pharmacopeia (USP) Reference Standard