Merck
  • Comparing various techniques to produce micro/nanoparticles for enhancing the dissolution of celecoxib containing PVP.
Comparing various techniques to produce micro/nanoparticles for enhancing the dissolution of celecoxib containing PVP.
European journal of pharmaceutics and biopharmaceutics : official journal of Arbeitsgemeinschaft fur Pharmazeutische Verfahrenstechnik e.V (2014-06-22)
Alireza Homayouni, Fatemeh Sadeghi, Jaleh Varshosaz, Hadi Afrasiabi Garekani, Ali Nokhodchi
ABSTRACT
One of the major challenges in pharmaceutical development is the poor dissolution performance of drugs. Celecoxib (CLX) is a poorly water soluble drug with its bioavailability being limited by its poor dissolution. In this study several particle engineering methods were employed on CLX using various ratios of CLX:PVP-K30. Micro/nanoparticles of CLX:PVP were prepared by using spray drying (SD), antisolvent crystallization followed by freeze drying (CRS-FD) and spray drying (CRS-SD) techniques. The suspension obtained through antisolvent crystallization was also subjected to high pressure homogenization followed by freeze drying (HPH-FD). Particle size measurements, saturation solubility, optical and scanning electron microscopy, DSC, XRPD, FT-IR and dissolution test were performed to characterize the physicochemical and pharmaceutical properties of the samples. The results showed that spray dried samples in the presence of (50%) PVP produced spherical particles and exhibited a high dissolution rate. Interestingly in the antisolvent crystallization technique, spherical nanoparticles of drug-PVP were obtained in the range of 291-442 nm. The average particle size was dependent on the concentration of the PVP used during the crystallization process. Solid state analysis showed that these particles were completely amorphous in nature. Also interesting to note was that at concentration of 5% PVP, when the suspension of nanoparticles was subjected to the high pressure homogenization process, the crystallinity of CLX increased. Despite the partial crystallinity of particles produced, they showed excellent dissolution behavior. It can thus be concluded that the method of preparation of CLX micro/nanoparticles had a big impact on the dissolution rate when the concentration of PVP was low (e.g., 5%). At high PVP concentration (e.g., 50%) all methods used to prepare engineered CLX particles showed better dissolution with no significant differences in their dissolution efficiency.
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, Molecular Biology, 10% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, 20% in H2O
Sodium laurilsulfate, European Pharmacopoeia (EP) Reference Standard
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Supelco
Sodium dodecyl sulfate, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, Molecular Biology, ≥98.5% (GC), free-flowing, Redi-Dri
Celecoxib, European Pharmacopoeia (EP) Reference Standard