Merck
  • Validated LC-MS/MS method for the simultaneous determination of chlorambucil and its prodrug in mouse plasma and brain, and application to pharmacokinetics.
Validated LC-MS/MS method for the simultaneous determination of chlorambucil and its prodrug in mouse plasma and brain, and application to pharmacokinetics.
Journal of pharmaceutical and biomedical analysis (2014-08-05)
XinYi Wang, Quan Zhang, Qing Lin, Yue Zhang, Zhi-Rong Zhang
ABSTRACT
A simple, sensitive and rapid LC-MS/MS method was developed and validated for the simultaneous determination of chlorambucil (CHL) and the prodrug of chlorambucil (CHLS) in mouse plasma and brain tissue. Detection was performed on a Diamonsil ODS chromatography column using gradient elution with a mobile phase of 0.2% aqueous formic acid and acetonitrile. Mass spectrometry was carried out in multiple reaction monitoring mode using a positive electrospray ionization interface. Good linearity was found for CHLS and CHL in plasma and brain tissue in different linear ranges (r>0.9996). Intra-day and inter-day precision was within 9.11% and accuracy was not more than 11.07%. The validated method was successfully applied to the pharmacokinetic study of CHLS and CHL in mice after intravenous administration.
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
USP
Residual Solvent Class 2 - Acetonitrile, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Supelco
Acetonitrile, analytical standard
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
Sigma-Aldrich
Chlorambucil
Supelco
Acetonitrile(Neat), Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Residual Solvent - Acetonitrile(solution in DMSO), Pharmaceutical Secondary Standard; Certified Reference Material
Chlorambucil for system suitability, European Pharmacopoeia (EP) Reference Standard
Chlorambucil, European Pharmacopoeia (EP) Reference Standard