Merck
  • Stability-indicating HPLC method for quantification of celecoxib and diacerein along with its impurities in capsule dosage form.
Stability-indicating HPLC method for quantification of celecoxib and diacerein along with its impurities in capsule dosage form.
Journal of chromatographic science (2014-05-20)
Hanimi Reddy Bapatu, Ravi Kumar Maram, R Satyanarayana Murthy
ABSTRACT
A novel stability-indicating reverse phase high performance liquid chromatography method was developed and validated for the simultaneous determination of Celecoxib (CEL) and Diacerein (DIN) and its impurities in capsule dosage form. The method was developed using L1 column with gradient using the mobile phase consist of Solution A (pH = 2.3 buffer) and Solution B (methanol and acetonitrile; 50 : 50, v/v). The eluted compounds were monitored at 255 nm. CEL and DIN were subjected to oxidative, acid, base, hydrolytic, thermal and photolytic stress conditions. The developed method was validated as per International Conference on Harmonisation guidelines with respect to specificity, linearity, limit of detection, limit of quantitation, accuracy, precision and robustness. The limit of quantitation results were ranged from 0.07 to 0.09 µg/mL for CEL impurities and 0.052 to 0.065 µg/mL for DIN impurities. This method is suitable for the estimation of impurities and assay of CEL and DIN in capsules dosage forms.
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Phosphoric acid solution, suitable for NMR (reference standard), 85% in D2O (99.9 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Phosphoric acid solution, suitable for NMR (reference standard), 85% in D2O (99.9 atom % D), NMR tube size 5 mm × 8 in.
Sigma-Aldrich
Triethylamine, ≥99%
Sigma-Aldrich
Triethylamine, ≥99.5%
Supelco
Methanol, analytical standard
Sigma-Aldrich
Triethylamine, puriss. p.a., ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, for amino acid analysis, ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, ampule, ≥99.5% (GC)
Sigma-Aldrich
Phosphoric acid solution, 85 wt. % in H2O, FCC, FG
Sigma-Aldrich
Triethylamine, ≥99.5%
Sigma-Aldrich
Phosphoric acid solution, suitable for NMR (reference standard), 85% in D2O (99.9 atom % D), NMR tube size 4.2 mm × 8 in. , WGS-5BL Coaxial NMR tube
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Triethylamine, analytical standard
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Supelco
Residual Solvent - Acetonitrile(solution in DMSO), Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Acetonitrile(Neat), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Phosphoric acid, crystalline, ≥99.999% trace metals basis
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%