Merck
  • lincRNA-RoR and miR-145 regulate invasion in triple-negative breast cancer via targeting ARF6.
lincRNA-RoR and miR-145 regulate invasion in triple-negative breast cancer via targeting ARF6.
Molecular cancer research : MCR (2014-09-26)
Gabriel Eades, Benjamin Wolfson, Yongshu Zhang, Qinglin Li, Yuan Yao, Qun Zhou
ABSTRACT
Triple-negative (ER(-), HER2(-), PR(-)) breast cancer (TNBC) is an aggressive disease with a poor prognosis with no available molecularly targeted therapy. Silencing of microRNA-145 (miR-145) may be a defining marker of TNBC based on molecular profiling and deep sequencing. Therefore, the molecular mechanism behind miR-145 downregulation in TNBC was examined. Overexpression of the long intergenic noncoding RNA regulator of reprogramming, lincRNA-RoR, functions as a competitive endogenous RNA sponge in TNBC. Interestingly, lincRNA-RoR is dramatically upregulated in TNBC and in metastatic disease and knockdown restores miR-145 expression. Previous reports suggest that miR-145 has growth-suppressive activity in some breast cancers; however, these data in TNBC indicate that miR-145 does not affect proliferation or apoptosis but instead, miR-145 regulates tumor cell invasion. Investigation of miR-145-regulated pathways involved in tumor invasion revealed a novel target, the small GTPase ADP-ribosylation factor 6 (Arf6). Subsequent analysis demonstrated that ARF6, a known regulator of breast tumor cell invasion, is dramatically upregulated in TNBC and in breast tumor metastasis. Mechanistically, ARF6 regulates E-cadherin localization and affects cell-cell adhesion. These results reveal a lincRNA-RoR/miR-145/ARF6 pathway that regulates invasion in TNBCs. The lincRNA-RoR/miR-145/ARF6 pathway is critical to TNBC metastasis and could serve as biomarkers or therapeutic targets for improving survival.
Product Description
USP
Hydrocortisone, United States Pharmacopeia (USP) Reference Standard
Supelco
Hydrocortisone, Pharmaceutical Secondary Standard; Certified Reference Material
Hydrocortisone, European Pharmacopoeia (EP) Reference Standard
Hydrocortisone, British Pharmacopoeia (BP) Assay Standard
Sigma-Aldrich
Hydrocortisone, ≥98% (HPLC)
Sigma-Aldrich
Anti-KI67 antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Hydrocortisone, BioReagent, suitable for cell culture
Sigma-Aldrich
Hydrocortisone, meets USP testing specifications
Sigma-Aldrich
Hydrocortisone, γ-irradiated, powder, BioXtra, suitable for cell culture
Sigma-Aldrich
Trimesic acid, 95%
Sigma-Aldrich
Trimethylaluminum, 97%
Sigma-Aldrich
Trimethylaluminum, packaged for use in deposition systems
Sigma-Aldrich
Trimethylaluminum solution, 2.0 M in heptane
Sigma-Aldrich
Trimethylaluminum solution, 2.0 M in toluene
Sigma-Aldrich
Trimethylaluminum solution, 2.0 M in hexanes
Hydrocortisone for peak identification, European Pharmacopoeia (EP) Reference Standard