Merck
  • Metrological traceability in mass spectrometry-based targeted protein quantitation: a proof-of-principle study for serum apolipoproteins A-I and B100.
Metrological traceability in mass spectrometry-based targeted protein quantitation: a proof-of-principle study for serum apolipoproteins A-I and B100.
Journal of proteomics (2014-06-28)
Nico P M Smit, Fred P H T M Romijn, Irene van den Broek, Jan W Drijfhout, Martin Haex, Arnoud van der Laarse, Yuri E M van der Burgt, Christa M Cobbaert
ABSTRACT
In this study, we have followed up on previous liquid chromatography (LC) multiple reaction monitoring (MRM) mass spectrometry (MS) approaches for measurement of apolipoprotein (apo) A-I and apo B100 in serum aiming for implementation of a multiplexed assay in a clinical chemistry laboratory with full metrological traceability. Signature peptides were selected and detected by dynamic MRM, and stable isotope labeled (SIL)-peptides were used as internal standards. Five apo A-I and four apo B100 peptides were measured in serum digests with linearity (R(2)>0.992) in the physiologically relevant concentration ranges. Linearity with regard to protein concentration was ascertained at five concentration levels (R(2)>0.926 and R(2)>0.965, for the apo A-I and apo B100 peptides, respectively). Three native value-assigned sera were used as external calibrators for further method verification. Imprecision values on sample preparation and LC-MS/MS acquisition were below the established minimal specifications for apo A-I and apo B100 (5.0% and 5.3%, respectively). Correlation of LC-MS/MS results with immunoturbidimetric assay results, for normo- and hypertriglyceridemic samples, showed R(2)>0.944 for apo A-I and R(2)>0.964 for apo B100. This LC-MS/MS method has potential for clinical application in normo- and dyslipidemic patients. Measurement of apo A-I and apo B100 may offer an alternative to high and low density lipoprotein cholesterol (HDL-c and LDL-c) methods for cardiovascular disease risk assessment in dyslipidemic patients [1]. An LC-MS/MS method for apo A-I and apo B100 has the advantage of antibody independent and specific detection of protein signature peptides. The introduction of an LC-MS/MS method for apo A-I and apo B100 can serve as an example for many existing and newly developed (multiplex) biomarker methods in quantitative clinical chemistry proteomics (qCCP). Such LC-MS/MS methods should meet basic clinical chemistry principles with regard to test evaluation [2]. Criteria for imprecision should be pre-defined, e.g., based on biological variation. The use of commutable and traceable serum-based calibrators will improve inter-laboratory reproducibility of LC-MS/MS methods and may contribute to a more rapid transition of biomarker discovery to clinical utility with benefit for the patient treatment and improvement of general health care.
Sigma-Aldrich
3,4,4′-Trichlorocarbanilide, 99%
Sigma-Aldrich
Ammonium bicarbonate, BioUltra, ≥99.5% (T)
Sigma-Aldrich
Indole-3-acetic acid sodium salt, suitable for plant cell culture, BioReagent, ≥98%
Sigma-Aldrich
Urea, BioXtra, pH 7.5-9.5 (20 °C, 5 M in H2O)
Sigma-Aldrich
Iodoacetamide, ≥99% (NMR), crystalline
Supelco
Urea, 8 M (after reconstitution with 16 mL high purity water)
Sigma-Aldrich
Methanol solution, suitable for NMR (reference standard), 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
β-D-Allose, rare aldohexose sugar
Sigma-Aldrich
Formic acid solution, BioUltra, 1.0 M in H2O
USP
Urea, United States Pharmacopeia (USP) Reference Standard
Urea, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Ammonium bicarbonate, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Formic acid, ACS reagent, ≥96%
Sigma-Aldrich
Formic acid, reagent grade, ≥95%
Sigma-Aldrich
Formic acid, ACS reagent, ≥88%
Supelco
Urea, analytical standard
SAFC
Iodoacetamide
USP
Methyl alcohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Iodoacetamide, BioUltra
Sigma-Aldrich
Urea, ReagentPlus®, ≥99.5%, pellets
Sigma-Aldrich
Iodoacetamide, Single use vial of 56 mg
Sigma-Aldrich
Urea, suitable for electrophoresis
Sigma-Aldrich
Urea, ACS reagent, 99.0-100.5%
Sigma-Aldrich
Urea, powder, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Urea, BioUltra, Molecular Biology, 99% (T)
Sigma-Aldrich
Urea, meets USP testing specifications
Sigma-Aldrich
Urea, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%, 99.0-101.0% (calc. on dry substance)
Sigma-Aldrich
Urea, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99%
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Millipore
Urea solution, suitable for microbiology, 40% in H2O