Merck
  • Estrogen increases Nrf2 activity through activation of the PI3K pathway in MCF-7 breast cancer cells.
Estrogen increases Nrf2 activity through activation of the PI3K pathway in MCF-7 breast cancer cells.
Experimental cell research (2014-08-31)
Juanjuan Wu, Devin Williams, Grant A Walter, Winston E Thompson, Neil Sidell
ABSTRACT
The actions of the transcription factor Nuclear factor erythroid 2-related factor (Nrf2) in breast cancer have been shown to include both pro-oncogenic and anti-oncogenic activities which is influenced, at least in part, by the hormonal environment. However, direct regulation of Nrf2 by steroid hormones (estrogen and progesterone) has received only scant attention. Nrf2 is known to be regulated by its cytosolic binding protein, Kelch-like ECH-associated protein 1 (Keap1), and by a Keap1-independent mechanism involving a series of phosphorylation steps mediated by phosphatidylinositol 3-kinase (PI3K) and glycogen synthase kinase 3 beta (GSK3β). Here, we report that estrogen (E2) increases Nrf2 activity in MCF7 breast cancer cells through activation of the PI3K/GSK3β pathway. Utilizing antioxidant response element (ARE)-containing luciferase reporter constructs as read-outs for Nrf2 activity, our data indicated that E2 increased ARE activity >14-fold and enhanced the action of the Nrf2 activators, tertiary butylhydroquinone (tBHQ) and sulforaphane (Sul) 4 to 9 fold compared with cells treated with tBHQ or Sul as single agents. This activity was shown to be an estrogen receptor-mediated phenomenon and was antagonized by progesterone. In addition to its action on the reporter constructs, mRNA and protein levels of heme oxygenase 1, an endogenous target gene of Nrf2, was markedly upregulated by E2 both alone and in combination with tBHQ. Importantly, E2-induced Nrf2 activation was completely suppressed by the PI3K inhibitors LY294002 and Wortmannin while the GSK3β inhibitor CT99021 upregulated Nrf2 activity. Confirmation that E2 was, at least partly, acting through the PI3K/GSK3β pathway was indicated by our finding that E2 increased the phosphorylation status of both GSK3β and Akt, a well-characterized downstream target of PI3K. Together, these results demonstrate a novel mechanism by which E2 can regulate Nrf2 activity in estrogen receptor-positive breast cancer cells and suggest that patients׳ hormonal status through this activity may play a significant role in some therapeutic outcomes.
Product Description
Medroxyprogesterone acetate, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Progesterone, powder, BioReagent, suitable for cell culture
Supelco
Progesterone, VETRANAL®, analytical standard
Supelco
Progesterone, Pharmaceutical Secondary Standard; Certified Reference Material
Progesterone for peak identification, European Pharmacopoeia (EP) Reference Standard
Progesterone, European Pharmacopoeia (EP) Reference Standard
USP
Progesterone, United States Pharmacopeia (USP) Reference Standard
Progesterone for system suitability, European Pharmacopoeia (EP) Reference Standard
Supelco
Medroxyprogesterone 17-acetate, VETRANAL®, analytical standard
Sigma-Aldrich
Medroxyprogesterone 17-acetate, ≥97% (HPLC)
USP
Medroxyprogesterone acetate, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Pyrimidine, ≥98.0%
Sigma-Aldrich
Progesterone, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
Progesterone, ≥99%
Sigma-Aldrich
tert-Butylhydroquinone, 97%
Sigma-Aldrich
Progesterone, meets USP testing specifications
Medroxyprogesterone acetate for peak identification, European Pharmacopoeia (EP) Reference Standard
Medroxyprogesterone acetate for system suitability, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
DL-Sulforaphane, ≥90% (HPLC), synthetic, liquid