Merck
Serotonin transporter polymorphism (5-HTTLPR) in Croatian population.
Molecular biology reports (2014-11-07)
Jelena Culej, Mario Štefanović, Ivana Ćelap, Nora Nikolac, Dalibor Karlović
ABSTRACT
Serotonin transporter polymorphism (5-HTTLPR) is a well-studied polymorphism in psychiatric research. The function of serotonin transporter is to control neural stimulation and maintain homeostasis of serotonin in other cells like platelets and enterochromaffin cells. Considering serotonin function in human behavior, and the role of serotonin transporter, 5-HTTLPR has been associated with depression related disorders, anxiety related personality traits, and adverse response to psychotherapy. However, many studies failed to replicate the association of 5-HTTLPR polymorphism with mentioned disorders. The aim of our study was to assess genotype frequencies in Croatian physically and psychologically healthy population and compare our results with previously published data. Genotype distribution in our research was similar to previous studies on Caucasian population regardless of inclusion criteria. Genotype distribution was as follows: LL 38 %; LS 45 %; SS 17 % and allele frequencies for L and S allele were 61 and 39 %, respectively. Obtained results were in an agreement with the Hardy-Weinberg equilibrium. Comparing inclusion criteria from different studies, we noticed a difference in population selection from one study to another. Increased possibility for selection bias, population stratification and complexity of psychiatric disorders might present a source of possible errors in genetic association studies.
Sigma-Aldrich
Magnesium chloride solution, 0.1 M
Sigma-Aldrich
Magnesium chloride solution, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Magnesium chloride solution, Molecular Biology, 1.00 M±0.01 M
Sigma-Aldrich
Magnesium chloride solution, PCR Reagent, 25 mM MgCI2 solution for PCR
Sigma-Aldrich
Magnesium chloride solution, BioUltra, Molecular Biology, ~0.025 M in H2O
Sigma-Aldrich
Magnesium chloride solution, BioUltra, Molecular Biology, 2 M in H2O
Sigma-Aldrich
Potassium hydride, in paraffin
Sigma-Aldrich
Magnesium chloride, suitable for insect cell culture, BioReagent, ≥97.0%
Sigma-Aldrich
Magnesium chloride, ≥98%
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.9% trace metals basis
Sigma-Aldrich
Potassium, chunks (in mineral oil), 98% trace metals basis
Sigma-Aldrich
Potassium hydride, 30 wt % dispersion in mineral oil
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Magnesium chloride, powder, <200 μm