Merck
  • Expanding functionality of recombinant human collagen through engineered non-native cysteines.
Expanding functionality of recombinant human collagen through engineered non-native cysteines.
Biomacromolecules (2014-08-22)
Richard Que, Ali Mohraz, Nancy A Da Silva, Szu-Wen Wang
ABSTRACT
Collagen is the most abundant protein in extracellular matrices and is commonly used as a tissue engineering scaffold. However, collagen and other biopolymers from native sources can exhibit limitations when tuning mechanical and biological properties. Cysteines do not naturally occur within the triple-helical region of any native collagen. We utilized a novel modular synthesis strategy to fabricate variants of recombinant human collagen that contained 2, 4, or 8 non-native cysteines at precisely defined locations within each biopolymer. This bottom-up approach introduced capabilities using sulfhydryl chemistry to form hydrogels and immobilize bioactive factors. Collagen variants retained their triple-helical structure and supported cellular adhesion. Hydrogels were characterized using rheology, and the storage moduli were comparable to fibrillar collagen gels at similar concentrations. Furthermore, the introduced cysteines functioned as anchoring sites, with TGF-β1-conjugated collagens promoting myofibroblast differentiation. This approach demonstrates the feasibility to produce custom-designed collagens with chemical functionality not available from native sources.
Product Description
Supelco
Acrylamide solution, 40% in H2O
Sigma-Aldrich
Nitrogen, ≥99.998%
Sigma-Aldrich
Acrylamide solution, 40%, suitable for electrophoresis, sterile-filtered
Sigma-Aldrich
4-(N-Maleimidomethyl)cyclohexanecarboxylic acid N-hydroxysuccinimide ester, ≥98%, powder
Millipore
D-(+)-Galactose, suitable for microbiology, ≥99.0%
Sigma-Aldrich
Acrylamide, suitable for electrophoresis, ≥99% (HPLC), powder
Sigma-Aldrich
Acrylamide, suitable for electrophoresis, ≥99%
Sigma-Aldrich
Acrylamide, Molecular Biology, ≥99% (HPLC)
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥98.5% (GC)
Sigma-Aldrich
D-(+)-Galactose, ≥99% (HPLC), BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
D-(+)-Galactose, ≥99% (HPLC)
Sigma-Aldrich
D-(+)-Galactose, ≥98% (HPLC)
Sigma-Aldrich
Acrylamide, purum, ≥98.0% (GC)
Sigma-Aldrich
D-(+)-Galactose, meets analytical specification of Ph. Eur., BP
Sigma-Aldrich
Acrylamide, for Northern and Southern blotting, powder blend
Sigma-Aldrich
D-(+)-Galactose, BioXtra, ≥99% (HPLC)
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥99.0% (T)
Galactose, European Pharmacopoeia (EP) Reference Standard
Supelco
Acrylamide, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
USP
Galactose, United States Pharmacopeia (USP) Reference Standard
Supelco
Acrylamide, analytical standard
Supelco
Galactose, Pharmaceutical Secondary Standard; Certified Reference Material