Merck
  • Phospholipid vesicle-based permeation assay and EpiSkin® in assessment of drug therapies destined for skin administration.
Phospholipid vesicle-based permeation assay and EpiSkin® in assessment of drug therapies destined for skin administration.
Journal of pharmaceutical sciences (2015-01-06)
André Engesland, Nataša Škalko-Basnet, Gøril Eide Flaten
ABSTRACT
Cost-effective and efficient methods for permeability screening are crucial during early development of drugs, drug formulations, and cosmeceuticals. Alternatives to animal experiments are impelled for both economical and ethical reasons. The aim of this study was to determine the ability of the phospholipid vesicle-based permeation assay (PVPA) to assess the effect of different formulations on drug permeability and thus establish its utility in formulation development. Three model drugs were tested in solutions and as liposomal formulations. The permeability results for the PVPA models were compared with the results for the reconstructed human skin model, EpiSkin(®). The drugs were ranked based on their estimated penetration potentials, and the results were in accordance with what was expected considering the physicochemical properties of the drugs. PVPAs (E-80, ceramide, cholesterol, cholesteryl sulfate, and palmitic acid) was able to distinguish between drug solutions and liposomal formulations; however, EpiSkin(®) detected only small differences between the drugs in solution and formulations. In contrast with EpiSkin(®), which is limited by a 3-day testing window, PVPA barriers can be stored frozen for up to 2 weeks or even up to 16 months, depending on their compositions. The PVPA models are thus more cost effective and efficient than the EpiSkin(®) model for permeability screening during early drug development.
Sigma-Aldrich
Chloroform, biotech. grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Aciclovir, European Pharmacopoeia (EP) Reference Standard
USP
Dehydrated Alcohol, United States Pharmacopeia (USP) Reference Standard
Supelco
Palmitic acid, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Palmitic acid, United States Pharmacopeia (USP) Reference Standard
Supelco
Cholesterol, Pharmaceutical Secondary Standard; Certified Reference Material
Palmitic acid, European Pharmacopoeia (EP) Reference Standard
Supelco
Acyclovir, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Supelco
Cholesterol solution, certified reference material, 10 mg/mL in chloroform
Sigma-Aldrich
SyntheChol® NS0 Supplement, 500 ×, synthetic cholesterol, animal component-free, sterile-filtered, aqueous solution, suitable for cell culture
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
Cholesterol, powder, BioReagent, suitable for cell culture, ≥99%
Sigma-Aldrich
Cholesterol, Sigma Grade, ≥99%
Sigma-Aldrich
Cholesterol, tested according to Ph. Eur.
Supelco
Hydrogen chloride – methanol solution, ~1.25 m HCl (T), derivatization grade (GC derivatization), LiChropur
Sigma-Aldrich
Cholesterol, from lanolin, ≥99.0% (GC)
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Supelco
Hydrogen chloride – 2-propanol solution, ~1.25 M HCl (T), derivatization grade (GC derivatization), LiChropur
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, Molecular Biology, 10 M in H2O
Sigma-Aldrich
Sodium hydroxide, pellets, semiconductor grade, 99.99% trace metals basis
Sigma-Aldrich
Sodium hydroxide, BioXtra, ≥98% (acidimetric), pellets (anhydrous)
Supelco
Sodium hydroxide solution, 49-51% in water, eluent for IC
Sigma-Aldrich
Sodium hydroxide solution, 50% in H2O
Sigma-Aldrich
Sodium hydroxide, reagent grade, 97%, flakes
Sigma-Aldrich
Sodium hydroxide, BioUltra, suitable for luminescence, ≥98.0% (T), pellets