Merck
  • Polo-like kinase 1 inhibitor BI2536 causes mitotic catastrophe following activation of the spindle assembly checkpoint in non-small cell lung cancer cells.
Polo-like kinase 1 inhibitor BI2536 causes mitotic catastrophe following activation of the spindle assembly checkpoint in non-small cell lung cancer cells.
Cancer letters (2014-12-20)
Minji Choi, Wootae Kim, Min Gyeong Cheon, Chang Woo Lee, Ja Eun Kim
ABSTRACT
Polo-like kinase 1 (PLK1), a critical kinase that regulates multiple steps in mitosis, is overexpressed in diverse human cancers; thus many PLK1 inhibitors have been developed as potential cancer therapeutic agents. One of these compounds, the PLK1-specific inhibitor BI2536, has been investigated as a cytotoxic drug in several cancers, including lung cancer; however, the detailed mechanism by which BI2536 induces defects in cell proliferation of non-small cell lung cancer (NSCLC) has not yet been determined. We found that BI2536 treatment resulted in mitotic arrest due to improper formation of the mitotic spindles and mitotic centrosomes. The unattached kinetochores in BI2536-treated NSCLC cells activated the spindle assembly checkpoint (SAC). The prolonged activation of the SAC led to a type of apoptotic cell death referred to as mitotic catastrophe. Finally, BI2536-treated NSCLC cells show a defect in cell proliferation. Overall, these data indicate that PLK1 inhibition via mitotic disruption represents a potential approach for the treatment of NSCLC.
Sigma-Aldrich
Sodium Fluoride Solution
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium fluoride 0.5 M solution
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Supelco
Fluoride ion solution for ISE, 0.1 M F-, for ion-selective electrodes
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, BioUltra, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, ≥99.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Sodium fluoride, 99.99% trace metals basis
Sigma-Aldrich
Sodium fluoride, ACS reagent, ≥99%
Sigma-Aldrich
Sodium fluoride, puriss., meets analytical specification of Ph. Eur., BP, USP, 98.5-100.5% (calc. to the dried substance)
Supelco
Sodium Fluoride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium fluoride, powder, 99.99% trace metals basis
Sigma-Aldrich
Sodium fluoride, BioXtra, ≥99%
Sigma-Aldrich
Sodium fluoride, BioReagent, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Sodium fluoride, ReagentPlus®, ≥99%