Merck
  • Characterization and functional studies of forkhead box protein 3(-) lymphocyte activation gene 3(+) CD4(+) regulatory T cells induced by mucosal B cells.
Characterization and functional studies of forkhead box protein 3(-) lymphocyte activation gene 3(+) CD4(+) regulatory T cells induced by mucosal B cells.
Clinical and experimental immunology (2015-01-13)
K-H Chu, B-L Chiang
ABSTRACT
The induction of mucosal tolerance has been demonstrated to be an effective therapeutic approach for the treatment of allergic diseases. Our previous study demonstrated that Peyer's patch B cells could convert naive T cells into regulatory T cells (so-called Treg -of-B(P) cells); however, it is important to characterize this particular subset of Treg -of-B cells for future applications. This study aimed to investigate the role of lymphocyte activating gene 3 (LAG3) in mediating the regulatory function of Treg -of-B(P) cells induced by mucosal follicular B (FOB) cells. Microarray analysis and real-time polymerase chain reaction (PCR) were used to assess the gene expression pattern of Treg -of-B(P) cells. To evaluate the role of LAG3, the in-vitro suppressive function and the alleviation of airway inflammation in a murine model of asthma was assessed. Our data indicated that FOB cells isolated from Peyer's patches had the ability to generate more suppressive Treg -of-B cells with LAG3 expression, compared with CD23(lo) CD21(lo) B cells. LAG3 is not only a marker for Treg -of-B(P) cells, but also participate in the suppressive ability. Moreover, CCR4 and CCR6 could be detected on the LAG3(+) , not LAG3(-) , Treg -of-B(P) cells and would help cells homing to allergic lung. In the murine model of asthma, the adoptive transfer of LAG3(+) Treg -of-B(P) cells was able to sufficiently suppress T helper type 2 (Th2) cytokine production, eosinophil infiltration and alleviate asthmatic symptoms. LAG3 was expressed in Treg -of-B(P) cells and was also involved in the function of Treg -of-B(P) cells. In the future, this particular subset of Treg -of-B cells might be used to alleviate allergic symptoms.
Sigma-Aldrich
Nitrogen, ≥99.998%
Sigma-Aldrich
Vinylene carbonate, contains ≤2% BHT as stabilizer, 97%