Merck
  • Efficient analysis of selected estrogens using fabric phase sorptive extraction and high performance liquid chromatography-fluorescence detection.
Efficient analysis of selected estrogens using fabric phase sorptive extraction and high performance liquid chromatography-fluorescence detection.
Journal of chromatography. A (2014-08-02)
Rajesh Kumar, Gaurav, Heena, Ashok Kumar Malik, Abuzar Kabir, Kenneth G Furton
ABSTRACT
A simple, fast and sensitive analytical method using fabric phase sorptive extraction (FPSE) followed by high performance liquid chromatography with fluorescence detection (HPLC-FLD) has been developed for efficient quantification of biologically important molecules e.g., 17α-ethynylestradiol (EE2), β-estradiol (E2) and bisphenol A (BPA). FPSE is a new sorptive extraction technique that integrates the advantages of permeable sol-gel derived hybrid organic-inorganic sorbents with flexible and permeable fabric substrates, resulting in a highly efficient and sensitive extraction media that can be introduced directly into any kind of fluidic matrix. Various factors affecting the performance of FPSE technique were optimized. The chromatographic separation was carried using mobile phase acetonitrile/methanol/water (30:15:55; v/v) at a flow rate 1.0mL/min on C18 column with fluorescence detection (λex=280nm and λem=310nm). The calibration curves of the target analytes were prepared with good correlation coefficient values (R(2)>0.992). Limit of detection (LOD) values range from 20 to 42pg/mL. The developed method was applied successfully for the analysis of estrogen molecules in urine and various kinds of aqueous samples.
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Dichloromethane, anhydrous, ≥99.8%, contains 40-150 ppm amylene as stabilizer
Supelco
Acetonitrile, analytical standard
Sigma-Aldrich
Dichloromethane, suitable for HPLC, ≥99.9%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Sodium hydroxide-16O solution, 20 wt. % in H216O, 99.9 atom % 16O
Supelco
Bisphenol A, ≥99%
Sigma-Aldrich
Bisphenol A, 97%
Supelco
Bisphenol A, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Methanol solution, suitable for NMR (reference standard), 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Supelco
Trifluoroacetic acid, analytical standard
Sigma-Aldrich
Dichloromethane, contains 40-150 ppm amylene as stabilizer, ACS reagent, ≥99.5%
Sigma-Aldrich
Trifluoroacetic acid, puriss. p.a., suitable for HPLC, ≥99.0% (GC)
Sigma-Aldrich
Trifluoroacetic acid, ReagentPlus®, 99%
Sigma-Aldrich
Dichloromethane, ACS reagent, ≥99.5%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Trifluoroacetic acid, ≥99%, for protein sequencing
Sigma-Aldrich
Dichloromethane, ACS reagent, ≥99.5%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Dichloromethane, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains 50-150 ppm amylene as stabilizer
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Supelco
Methylene Chloride, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Methyl alcohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
Sigma-Aldrich
Trifluoroacetic acid, suitable for HPLC, ≥99.0%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Dichloromethane, suitable for HPLC, ≥99.8%, contains amylene as stabilizer
USP
Residual Solvent Class 2 - Acetonitrile, United States Pharmacopeia (USP) Reference Standard
Supelco
Dichloromethane, analytical standard