Merck
  • Single-walled carbon nanotube-based polymer monoliths for the enantioselective nano-liquid chromatographic separation of racemic pharmaceuticals.
Single-walled carbon nanotube-based polymer monoliths for the enantioselective nano-liquid chromatographic separation of racemic pharmaceuticals.
Journal of chromatography. A (2014-08-19)
Marwa Ahmed, Mir Massoud Aghili Yajadda, Zhao Jun Han, Dawei Su, Guoxiu Wang, Kostya Ken Ostrikov, Ashraf Ghanem
ABSTRACT
Single-walled carbon nanotubes were encapsulated into different polymer-based monolithic backbones. The polymer monoliths were prepared via the copolymerization of 20% monomers, glycidyl methacrylate, 20% ethylene glycol dimethacrylate and 60% porogens (36% 1-propanol, 18% 1,4-butanediol) or 16.4% monomers (16% butyl methacrylate, 0.4% sulfopropyl methacrylate), 23.6% ethylene glycol dimethacrylate and 60% porogens (36% 1-propanol, 18% 1,4-butanediol) along with 6% single-walled carbon nanotubes aqueous suspension. The effect of single-walled carbon nanotubes on the chiral separation of twelve classes of pharmaceutical racemates namely; α- and β-blockers, antiinflammatory drugs, antifungal drugs, dopamine antagonists, norepinephrine-dopamine reuptake inhibitors, catecholamines, sedative hypnotics, diuretics, antihistaminics, anticancer drugs and antiarrhythmic drugs was investigated. The enantioselective separation was carried out under multimodal elution to explore the chiral recognition capabilities of single-walled carbon nanotubes using reversed phase, polar organic and normal phase chromatographic conditions using nano-liquid chromatography. Baseline separation was achieved for celiprolol, chlorpheniramine, etozoline, nomifensine and sulconazole under multimodal elution conditions. Satisfactory repeatability was achieved through run-to-run, column-to-column and batch-to-batch investigations. Our findings demonstrate that single-walled carbon nanotubes represent a promising stationary phase for the chiral separation and may open the field for a new class of chiral selectors.
Product Description
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Acetone, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Acetone, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
1-Propanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Acetone, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Ethyl alcohol, Pure 190 proof, for molecular biology
Sigma-Aldrich
Formamide, spectrophotometric grade, ≥99%
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Sigma-Aldrich
Formamide, ReagentPlus®, ≥99.0% (GC)
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Acetone, histological grade, ≥99.5%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Supelco
1-Propanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Glycidyl methacrylate, ≥97.0% (GC)
Sigma-Aldrich
Ethylene glycol dimethacrylate, 98%, contains 90-110 ppm monomethyl ether hydroquinone as inhibitor
Sigma-Aldrich
Acetone, natural, ≥97%
Sigma-Aldrich
Acetone, ≥99%, FCC, FG
Supelco
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Formamide, ≥99.5% (GC), BioReagent, Molecular Biology
Sigma-Aldrich
Formamide, BioUltra, Molecular Biology, ≥99.5% (T)
Supelco
Acetone, analytical standard