Merck
  • LPS stimulates and Hsp70 down-regulates TLR4 to orchestrate differential cytokine response of culture-differentiated innate memory CD8(+) T cells.
LPS stimulates and Hsp70 down-regulates TLR4 to orchestrate differential cytokine response of culture-differentiated innate memory CD8(+) T cells.
Cytokine (2015-02-24)
Amlan Kanti Ghosh, Debolina Sinha, Subhadeep Mukherjee, Ratna Biswas, Tapas Biswas
ABSTRACT
Nonconventional innate memory CD8(+) T cells characteristically expressing CD44, CD122, eomesodermin (Eomes) and promyelocytic leukemia zinc finger (PLZF) were derived in culture from CD4(+)CD8(+) double positive (DP) thymocytes of normal BALB/c and C57BL/6 mice. These culture-differentiated cells constitutively express toll-like receptor (TLR)4 and release interferon (IFN)-γ and interleukin (IL)-10. We show the TLR4-ligand lipopolysaccharide (LPS) stimulate the TLR and up-regulate IFN-γ skewing the cells towards type 1 polarization. In presence of LPS these cells also express suppressor of cytokine signaling (SOCS)1 and thus suppress IL-10 expression. In contrast, heat shock protein (Hsp)70 down-regulated TLR4 augmenting the anti-inflammatory cytokine IL-10. In association with IL-10 release IFN-γ was abrogated. The programmed cell death (PD)-1 mostly present in regulatory T cells was stimulated in these IL-10 producing cells by Hsp70 and not LPS indicating the cells can be driven to two contrast outcomes by the two TLR4 ligands. Our work provides a scope for in vitro monitoring of CD8(+) T cells to decipher important immune therapeutic option during infection or sepsis.
Product Description
Supelco
Aphidicolin, analytical standard
Sigma-Aldrich
Aphidicolin from Nigrospora sphaerica, ≥98% (HPLC), powder
Sigma-Aldrich
Selenium, powder, −100 mesh, ≥99.5% trace metals basis
Sigma-Aldrich
Selenium, powder, −100 mesh, 99.99% trace metals basis
Sigma-Aldrich
Selenium, pellets, <5 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Selenium, pellets, <5 mm particle size, ≥99.999% trace metals basis
Selenium, foil, 25x25mm, thickness 3mm, 99.95%
Selenium, pellets, < 5mm, ≥99.999%