Merck
  • Retinoids and rexinoids inhibit hepatitis C virus independently of retinoid receptor signaling.
Retinoids and rexinoids inhibit hepatitis C virus independently of retinoid receptor signaling.
Microbes and infection (2013-11-02)
Yuko Murakami, Masayoshi Fukasawa, Yukihiro Kaneko, Tetsuro Suzuki, Takaji Wakita, Hidesuke Fukazawa
ABSTRACT
Using a high-throughput screening system involving HCV JFH-1-Huh 7.5.1 cells, we determined that the ligands of class II nuclear receptors, retinoids and rexinoids inhibit HCV infection. Retinoids, ligands of retinoic acid receptor (RAR), and rexinoids, ligands of retinoid X receptor (RXR), reduced extracellular HCV RNA of HCV infected cells in a dose-dependent manner. The 50% effective concentrations were below 10 nM, and the 50% cytotoxic concentrations were over 10 μM. Both agonists and antagonists demonstrated inhibition, which indicates that the effect is not dependent on retinoic acid signaling. These chemicals reduced HCV RNA and NS5A protein levels in cells harboring the subgenomic HCV replicon RNA, which suggests that the chemicals affect HCV RNA replication. These compounds were also effective against persistently infected cells, although the reduction in the intracellular HCV RNA was smaller than that of the extracellular HCV RNA, suggesting that viral post-replication step is also inhibited. In combination with interferon (IFN), retinoid exhibited a synergistic effect. Retinoids did not enhance expression of the IFN effector molecule PKR. These series of compounds warrant further investigation as new class of HCV drugs, for the clinical translation of our observation may lead to increased anti-HCV efficacy.
Product Description
Supelco
Tretinoin, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Tretinoin, United States Pharmacopeia (USP) Reference Standard
Tretinoin, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
L-Thyroxine, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
L-Thyroxine, ≥98% (HPLC)
Sigma-Aldrich
Cholecalciferol, ≥98% (HPLC)
Sigma-Aldrich
SR11237, ≥98% (HPLC)
Cholecalciferol, European Pharmacopoeia (EP) Reference Standard
USP
Cholecalciferol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Cholecalciferol, analytical standard
Sigma-Aldrich
Cholecalciferol, meets USP testing specifications
Supelco
Cholecalciferol (D3), analytical standard
Sigma-Aldrich
Vitamin D3 solution, 100 μg/mL in ethanol, 97% (CP)
Adapalene, European Pharmacopoeia (EP) Reference Standard
Clofibrate, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Clofibrate, liquid
Supelco
Vitamin D3 solution, 1 mg/mL in ethanol, ampule of 1 mL, certified reference material, Cerilliant®
Supelco
Cholecalciferol (Vitamin D3), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Adapalene, ≥98% (HPLC)
Sigma-Aldrich
Bexarotene, ≥98% (HPLC)
USP
Levothyroxine, United States Pharmacopeia (USP) Reference Standard
USP
Adapalene, United States Pharmacopeia (USP) Reference Standard
Supelco
Adapalene, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Clofibrate, analytical standard