Merck
  • Highly flexible and rapidly degradable papaverine-loaded electrospun fibrous membranes for preventing vasospasm and repairing vascular tissue.
Highly flexible and rapidly degradable papaverine-loaded electrospun fibrous membranes for preventing vasospasm and repairing vascular tissue.
Acta biomaterialia (2014-04-08)
Wankun Zhu, Shenghe Liu, Jingwen Zhao, Shen Liu, Shichao Jiang, Bin Li, Huilin Yang, Cunyi Fan, Wenguo Cui
ABSTRACT
Vasospasm is a common post-operative complication after vascular anastomosis. Currently, the main treatment is a local injection of antispasmodic drugs. However, this method has a high rate of relapse and is subject to a large degree of individual variation, and repeated injections cause additional pain for patients. In this study, we developed highly flexible and rapidly degradable papaverine-loaded electrospun fibrous membranes to be wrapped around vascular suturing to prevent vasospasm. Poly-l-lactic acid/polyethylene glycol (PLLA/PEG) electrospun fibers containing papaverine maintained a high degree of flexibility and could withstand any folding, and are therefore suitable for wrapping vascular suturing. A rapid release of papaverine, between 2 and 7 days, was achieved by adjusting the proportions of PEG and PLLA. PLLA electrospun fibers containing 40% PEG (PLLA-40%) could control drug release and polymer degradation most effectively during the first 2 weeks post-operation. Testing using an in vivo rabbit model showed that PLLA-40% fibrous membranes produced significant antispasmodic effect without observable inflammation or hyperplasia, and the fibrous membranes were ideally biodegradable, with no impact on regional blood flow, pressure, vessel diameter or surrounding tissue hyperplasia. Therefore, papaverine-loaded electrospun fibrous membranes show the potential to greatly reduce post-operative vasospasm and maintain regular vascular morphology during antispasmodic therapy.
Product Description
Papaverine hydrochloride, European Pharmacopoeia (EP) Reference Standard
Supelco
Papaverine hydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
1,1,1,3,3,3-Hexafluoro-2-propanol, ≥99%
Supelco
1,1,1,3,3,3-Hexafluoro-2-propanol, derivatization grade (GC derivatization), LiChropur, ≥99.8%
Sigma-Aldrich
Papaverine hydrochloride, powder