Merck
  • Profound prevention of experimental brain metastases of breast cancer by temozolomide in an MGMT-dependent manner.
Profound prevention of experimental brain metastases of breast cancer by temozolomide in an MGMT-dependent manner.
Clinical cancer research : an official journal of the American Association for Cancer Research (2014-03-19)
Diane Palmieri, Renata Duchnowska, Stephan Woditschka, Emily Hua, Yongzhen Qian, Wojciech Biernat, Katarzyna Sosińska-Mielcarek, Brunilde Gril, Andreas M Stark, Stephen M Hewitt, David J Liewehr, Seth M Steinberg, Jacek Jassem, Patricia S Steeg
ABSTRACT
Brain metastases of breast cancer cause neurocognitive damage and are incurable. We evaluated a role for temozolomide in the prevention of brain metastases of breast cancer in experimental brain metastasis models. Temozolomide was administered in mice following earlier injection of brain-tropic HER2-positive JIMT-1-BR3 and triple-negative 231-BR-EGFP sublines, the latter with and without expression of O(6)-methylguanine-DNA methyltransferase (MGMT). In addition, the percentage of MGMT-positive tumor cells in 62 patient-matched sets of breast cancer primary tumors and resected brain metastases was determined immunohistochemically. Temozolomide, when dosed at 50, 25, 10, or 5 mg/kg, 5 days per week, beginning 3 days after inoculation, completely prevented the formation of experimental brain metastases from MGMT-negative 231-BR-EGFP cells. At a 1 mg/kg dose, temozolomide prevented 68% of large brain metastases, and was ineffective at a dose of 0.5 mg/kg. When the 50 mg/kg dose was administered beginning on days 18 or 24, temozolomide efficacy was reduced or absent. Temozolomide was ineffective at preventing brain metastases in MGMT-transduced 231-BR-EGFP and MGMT-expressing JIMT-1-BR3 sublines. In 62 patient-matched sets of primary breast tumors and resected brain metastases, 43.5% of the specimens had concordant low MGMT expression, whereas in another 14.5% of sets high MGMT staining in the primary tumor corresponded with low staining in the brain metastasis. Temozolomide profoundly prevented the outgrowth of experimental brain metastases of breast cancer in an MGMT-dependent manner. These data provide compelling rationale for investigating the preventive efficacy of temozolomide in a clinical setting.
Sigma-Aldrich
Temozolomide, ≥98% (HPLC)
Supelco
Temozolomide, VETRANAL®, analytical standard
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Mgmt
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
L-Glutamine
Supelco
L-Glutamine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture
SAFC
L-Glutamine
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Supelco
L-Glutamine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Temozolomide, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
MISSION® esiRNA, targeting human MGMT