Merck
  • Antitumor activity of the ERK inhibitor SCH772984 [corrected] against BRAF mutant, NRAS mutant and wild-type melanoma.
Antitumor activity of the ERK inhibitor SCH772984 [corrected] against BRAF mutant, NRAS mutant and wild-type melanoma.
Molecular cancer (2014-08-22)
Deborah J L Wong, Lidia Robert, Mohammad S Atefi, Amanda Lassen, Geetha Avarappatt, Michael Cerniglia, Earl Avramis, Jennifer Tsoi, David Foulad, Thomas G Graeber, Begonya Comin-Anduix, Ahmed Samatar, Roger S Lo, Antoni Ribas
ABSTRACT
In melanoma, dysregulation of the MAPK pathway, usually via BRAF(V600) or NRAS(Q61) somatic mutations, leads to constitutive ERK signaling. While BRAF inhibitors are initially effective for BRAF-mutant melanoma, no FDA-approved targeted therapies exist for BRAF-inhibitor-resistant BRAF(V600), NRAS mutant, or wild-type melanoma. The 50% inhibitory concentration (IC50) of SCH772984, a novel inhibitor of ERK1/2, was determined in a panel of 50 melanoma cell lines. Effects on MAPK and AKT signaling by western blotting and cell cycle by flow cytometry were determined. Sensitivity fell into three groups: sensitive, 50% inhibitory concentration (IC50) < 1 μM; intermediately sensitive, IC50 1-2 μM; and resistant, >2 μM. Fifteen of 21 (71%) BRAF mutants, including 4 with innate vemurafenib resistance, were sensitive to SCH772984. All three (100%) BRAF/NRAS double mutants, 11 of 14 (78%) NRAS mutants and 5 of 7 (71%) wild-type melanomas were sensitive. Among BRAF(V600) mutants with in vitro acquired resistance to vemurafenib, those with MAPK pathway reactivation as the mechanism of resistance were sensitive to SCH772984. SCH772984 caused G1 arrest and induced apoptosis. Combining vemurafenib and SCH722984 in BRAF mutant melanoma was synergistic in a majority of cell lines and significantly delayed the onset of acquired resistance in long term in vitro assays. Therefore, SCH772984 may be clinically applicable as a treatment for non-BRAF mutant melanoma or in BRAF-mutant melanoma with innate or acquired resistance, alone or in combination with BRAF inhibitors.
Product Description
Supelco
L-Glutamine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
SAFC
L-Glutamine
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
β-D-Allose, rare aldohexose sugar
Sigma-Aldrich
L-Glutamine
Supelco
L-Glutamine, Pharmaceutical Secondary Standard; Certified Reference Material