Merck
  • Endogenous enzymes involved in the transformation of oleuropein in Spanish table olive varieties.
Endogenous enzymes involved in the transformation of oleuropein in Spanish table olive varieties.
Journal of agricultural and food chemistry (2014-09-12)
Eva Ramírez, Eduardo Medina, Manuel Brenes, Concepción Romero
ABSTRACT
The main Spanish table olive varieties supplied by different olive cooperatives were investigated for their polyphenol compositions and the endogenous enzymes involved in their transformations during two growing seasons. Olives of the Manzanilla variety had the highest concentration in total polyphenols, followed by the Hojiblanca and Gordal varieties. The Gordal and Manzanilla cultivars showed the highest polyphenol oxidase activities. The Gordal cultivar presented a greater β-glucosidase and esterase activity than the others. An important influence of pH and temperature on the optimal activity of these enzymes was also observed. The polyphenol oxidase activity increased with temperature, and peroxidase activity was optimal at 35 °C. The β-glucosidase and esterase activities were at their maximum at 30 and 55 °C, respectively. The oxidase and β-glucosidase activities were at their maximum at the pH of the raw fruit. These results will contribute to the knowledge of the enzyme transformation of oleuropein in natural table olives.
Sigma-Aldrich
2-(4-Hydroxyphenyl)ethanol, 98%
Sigma-Aldrich
4-Nitrophenol solution, 10 mM
Supelco
Guaiacol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Guaiacol, natural, ≥99%, FG
Supelco
2-(4-Hydroxyphenyl)ethanol, analytical standard
Sigma-Aldrich
Apigenin, ≥95.0% (HPLC)
Supelco
4-Nitrophenol, PESTANAL®, analytical standard
Sigma-Aldrich
4-Nitrophenyl acetate, esterase substrate
Sigma-Aldrich
Guaiacol, oxidation indicator
Sigma-Aldrich
4-Nitrophenol, spectrophotometric grade
Sigma-Aldrich
p-Coumaric acid, ≥98.0% (HPLC)
Supelco
Acetaminophen Related Compound F, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Apigenin, analytical standard
Guaiacol, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
4-Nitrophenol, ReagentPlus®, ≥99%
Supelco
Luteolin 7-glucoside, analytical standard
Supelco
trans-p-Coumaric acid, analytical standard
Sigma-Aldrich
Caffeic acid, ≥98.0% (HPLC)
Sigma-Aldrich
Luteolin, ≥98% (TLC), powder
Sigma-Aldrich
Luteolin 7-O-β-D-glucoside, ≥98.0% (HPLC)
Supelco
Luteolin, analytical standard
Supelco
Caffeic acid, suitable for matrix substance for MALDI-MS, ≥99.0% (HPLC)
Luteolin, primary reference standard
Luteolin 7-glucoside, primary reference standard