Merck
  • Placenta growth factor and vascular endothelial growth factor a have differential, cell-type specific patterns of expression in vascular cells.
Placenta growth factor and vascular endothelial growth factor a have differential, cell-type specific patterns of expression in vascular cells.
Microcirculation (New York, N.Y. : 1994) (2014-01-15)
Lingjin Xiang, Rohan Varshney, Nabil A Rashdan, Jennifer H Shaw, Pamela G Lloyd
ABSTRACT
PLGF, a VEGF-A related protein, mediates collateral enlargement via monocytes but plays little role in capillary proliferation. In contrast, VEGF-A mediates both collateral enlargement and capillary proliferation. PLGF has been less thoroughly studied than VEGF-A, and questions remain regarding its regulation and function. Therefore, our goal was to characterize the expression of PLGF by vascular cells. We hypothesized that vascular SMC would express more PLGF than EC, since VEGF-A is primarily expressed by non-EC. We compared PLGF and VEGF-A across eight EC and SMC lines, then knocked down PLGF and evaluated cell function. We also assessed the effect of hypoxia on PLGF expression and promoter activity. PLGF was most highly expressed in EC, whereas VEGF-A was most highly expressed in SMC. PLGF knockdown did not affect EC number, migration, or tube formation, but reduced monocyte migration toward EC. Monocyte migration was rescued by exogenous PLGF. Hypoxia increased PLGF protein without activating PLGF gene transcription. PLGF and VEGF-A have distinct patterns of expression in vascular cells. EC derived PLGF may function primarily in communication between EC and circulating cells. Hypoxia increases EC PLGF expression posttranscriptionally.
Sigma-Aldrich
Cobalt(II) chloride, AnhydroBeads, −10 mesh, 99.995% trace metals basis
Sigma-Aldrich
Calcein-AM, suitable for fluorescence, BioReagent, ≥90% (HPLC)
Sigma-Aldrich
Cobalt(II) chloride, AnhydroBeads, −10 mesh, 99.9% trace metals basis
Sigma-Aldrich
Calcein-AM, Small Package (20 X 50 μg ), ≥90.0% (HPLC)
Sigma-Aldrich
Cobalt(II) chloride, 97%
Sigma-Aldrich
Cobalt(II) chloride, purum p.a., anhydrous, ≥98.0% (KT)
Sigma-Aldrich
Mouse VEGF ELISA Kit, for serum, plasma, cell culture supernatant
Sigma-Aldrich
Human VEGF ELISA Kit, for cell and tissue lysates
Sigma-Aldrich
Mouse VEGF ELISA Kit, for cell and tissue lysates
Sigma-Aldrich
Rat VEGF ELISA Kit, for cell and tissue lysates
Sigma-Aldrich
Rat VEGF ELISA Kit, for serum, plasma and cell culture supernatant
Sigma-Aldrich
Human VEGF R2 ELISA Kit, for serum, plasma, cell culture supernatant and urine
Sigma-Aldrich
Human VEGF R3 ELISA Kit, for serum, plasma, cell culture supernatant, urine
Sigma-Aldrich
Human VEGF-A ELISA Kit, for serum, plasma, cell culture supernatants and urine
Sigma-Aldrich
Placenta Growth Factor human, PlGF, recombinant, expressed in E. coli, powder, suitable for cell culture
Sigma-Aldrich
Calcein AM solution, 4 mM in DMSO, ≥90% (HPLC), solution