Merck
  • MicroRNA-19b functions as potential anti-thrombotic protector in patients with unstable angina by targeting tissue factor.
MicroRNA-19b functions as potential anti-thrombotic protector in patients with unstable angina by targeting tissue factor.
Journal of molecular and cellular cardiology (2014-07-08)
Sufang Li, Jingyi Ren, Ning Xu, Jing Zhang, Qiang Geng, Chengfu Cao, Chongyou Lee, Junxian Song, Jingjin Li, Hong Chen
ABSTRACT
The activation of a hemostatic system plays a critical role in the incidence of acute coronary events. Hemostatic proteins may be regulated by microRNAs (miRNAs). Microparticles (MPs) are the major carrier of circulating miRNAs. The aim of this study was to determine the potential role of miRNAs in regulating gene expression involved in the hemostatic system in patients with unstable angina (UA). MiRNA expression profiles in the plasma from patients with UA (UA group, n=9) compared with individuals with clinical suspicion of coronary artery disease (CAD) but negative angiography (control group, n=9) showed that among 36 differentially expressed miRNAs, miR-19b was the most obvious one. Using real-time PCR, 5 selected miRNA levels in plasma (UA group, n=20; control group, n=30) and plasma MPs (UA group n=6; control group n=6) were proved to be consistent with the miRNA array. Flow cytometry analysis indicated that the amounts of plasma endothelial microparticles (EMPs) were increased in UA patients (UA group, n=4) compared to controls (control group, n=4). In cultured endothelial cells (ECs), TNF-α increased miR-19b release and expression. Tissue factor (TF) was predicted to be the target of miR-19b by bioinformatics analysis. Luciferase reporter assays demonstrated that miR-19b binds to TF mRNA. Overexpression of miR-19b inhibited TF expression and procoagulant activity. This study indicates that in UA patients, the increase of miR-19b wrapped in EMPs due to endothelial dysfunction may partially contribute to the circulating miR-19b elevation and miR-19b may play an anti-thrombotic role by inhibiting the expression of TF in ECs.
Sigma-Aldrich
TNF-α human, Animal-component free, recombinant, expressed in E. coli, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, ≥99.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, BioUltra, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Tumor Necrosis Factor-α human, TNF-α, recombinant, expressed in E. coli, powder, suitable for cell culture
Sigma-Aldrich
Tumor Necrosis Factor-α human, Xeno-free, recombinant, expressed in HEK 293 cells, suitable for cell culture