Merck
  • Nectin-4 expression contributes to tumor proliferation, angiogenesis and patient prognosis in human pancreatic cancer.
Nectin-4 expression contributes to tumor proliferation, angiogenesis and patient prognosis in human pancreatic cancer.
Journal of experimental & clinical cancer research : CR (2015-04-19)
Satoshi Nishiwada, Masayuki Sho, Satoshi Yasuda, Keiji Shimada, Ichiro Yamato, Takahiro Akahori, Shoichi Kinoshita, Minako Nagai, Noboru Konishi, Yoshiyuki Nakajima
ABSTRACT
Nectin-4 belongs to the nectin family that has diverse physiological and pathological functions in humans. Recent studies have also suggested some roles for Nectin-4 in several human cancers. However, the precise roles and clinical relevance of Nectin-4 in tumors are largely unknown. Nectin-4 expression was investigated in 123 patients with pancreatic cancer by immunohistochemistry. Furthermore, we investigated the association of Nectin-4 in pancreatic cancer with tumor proliferation, angiogenesis and immunity by using immunohistochemistry and siRNA interference method. Patients with high Nectin-4 expression had poorer postoperative prognosis than those with low expression. Importantly, multivariate analysis indicated that Nectin-4 expression had a significant independent prognostic value in pancreatic cancer (HR = 1.721, 1.085-2.730; P = 0.021). Tumor Nectin-4 expression was significantly correlated with Ki67 expression. In addition, siRNA-mediated gene silencing of Nectin-4 significantly inhibited the cell proliferation in human pancreatic cancer cells, Capan-2 and BxPC-3. Furthermore, Nectin-4 expression was also positively correlated with VEGF expression and intratumoral microvessel density. However, there were no significant correlations of tumor Nectin-4 expression with tumor-infiltrating T cells. Nectin-4 is a significant prognostic predictor, and may play a critical role in pancreatic cancer. Nectin-4 may be novel therapeutic target for pancreatic cancer.
Product Description
Supelco
Ethanol, standard for GC
Sigma-Aldrich
Ethanol, tested according to Ph. Eur.
Sigma-Aldrich
Ethyl alcohol, Pure 200 proof, Molecular Biology
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Ethanol, BioUltra, Molecular Biology, ≥99.8%, (absolute alcohol, without additive, A15 o1)
Sigma-Aldrich
Ethanol
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
Sigma-Aldrich
Ethanol, purum, fine spirit, denaturated with 4.8% methanol, F25 METHYL1, ~96% (based on denaturant-free substance)
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
MISSION® esiRNA, targeting human NECTIN4
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
Ethyl alcohol, Pure 190 proof, for molecular biology
Supelco
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Dehydrated Alcohol, United States Pharmacopeia (USP) Reference Standard