Merck
  • Peritoneal cavity is a route for gut-derived microbial signals to promote autoimmunity in non-obese diabetic mice.
Peritoneal cavity is a route for gut-derived microbial signals to promote autoimmunity in non-obese diabetic mice.
Scandinavian journal of immunology (2014-11-21)
R Emani, C Alam, S Pekkala, S Zafar, M R Emani, A Hänninen
ABSTRACT
Macrophages play a crucial role in innate immune reactions, and peritoneal macrophages (PMs) guard the sterility of this compartment mainly against microbial threat from the gut. Type 1 diabetes (T1D) is an autoimmune disease in which gut microbiota and gut immune system appear to contribute to disease pathogenesis. We have recently reported elevated free radical production and increased permeability of gut epithelium in non-obese diabetic (NOD) mice. Impaired barrier function could lead to bacterial leakage to the peritoneal cavity. To explore the consequences of impaired gut barrier function on extra-intestinal immune regulation, we characterized peritoneal lavage cells from young newly weaned NOD mice. We detected a rapid increase in the number of macrophages 1-2 weeks after weaning in NOD mice compared to C57BL/6 and BALB/c mice. Interestingly, this increase in macrophages was abrogated in NOD mice that were fed an antidiabetogenic diet (ProSobee), which improves gut barrier function. Macrophages in young (5-week-old) NOD mice displayed a poor TNF-α cytokine response to LPS stimulation and high expression of interleukin-1receptor-associated kinase-M (IRAK-M), indicating prior in vivo exposure to TLR-4 ligand(s). Furthermore, injection of LPS intraperitoneally increased T cell CD69 expression in pancreatic lymph node (PaLN), suggestive of T cell activation. Leakage of bacterial components such as endotoxins into the peritoneal cavity may contribute to auto-reactive T cell activation in the PaLN.
Sigma-Aldrich
Glycerol, ReagentPlus®, ≥99.0% (GC)
SAFC
L-Glutamine
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, Molecular Biology, 10% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, 20% in H2O
Sigma-Aldrich
Bromophenol Blue, titration: suitable
Sigma-Aldrich
Bromophenol Blue, ACS reagent
Supelco
L-Glutamine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture
Supelco
L-Glutamine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-Glutamine
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Supelco
Glycerol, analytical standard
Sigma-Aldrich
Glycerin, meets USP testing specifications
Sigma-Aldrich
Glycerol, Molecular Biology, ≥99.0%
Sigma-Aldrich
Glycerol, BioUltra, Molecular Biology, anhydrous, ≥99.5% (GC)
Sigma-Aldrich
Glycerol, BioXtra, ≥99% (GC)
Sigma-Aldrich
Glycerol, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
Sigma-Aldrich
Glycerol, ≥99.5%
Sigma-Aldrich
Glycerol, tested according to Ph. Eur., anhydrous
Sigma-Aldrich
Glycerol, ACS reagent, ≥99.5%
Sigma-Aldrich
Glycerol, FCC, FG
Sodium laurilsulfate, European Pharmacopoeia (EP) Reference Standard
Supelco
Glycerin, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Glycerin, United States Pharmacopeia (USP) Reference Standard
Supelco
Aphidicolin, analytical standard
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Aphidicolin from Nigrospora sphaerica, ≥98% (HPLC), powder
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, Molecular Biology, ≥98.5% (GC), free-flowing, Redi-Dri