Merck
  • Green synthesis, characterization and anti-inflammatory activity of silver nanoparticles using European black elderberry fruits extract.
Green synthesis, characterization and anti-inflammatory activity of silver nanoparticles using European black elderberry fruits extract.
Colloids and surfaces. B, Biointerfaces (2014-09-02)
Luminita David, Bianca Moldovan, Adriana Vulcu, Liliana Olenic, Maria Perde-Schrepler, Eva Fischer-Fodor, Adrian Florea, Maria Crisan, Ioana Chiorean, Simona Clichici, Gabriela Adriana Filip
ABSTRACT
This research aimed at reporting the synthesis, characterization and evaluation of the anti-inflammatory effects of some new biomaterials based on silver nanoparticles and polyphenols rich natural extracts. A fast and eco-friendly extracellular biosynthesis of silver nanoparticles (AgNPs), using European black elderberry (Sambucus nigra - SN, Adoxaceae family) fruit extracts was developed. The phytosynthesized nanoparticles exhibited an absorbance peak at 426nm, characteristic for AgNPs and their sizes were ranged from 20 to 80nm. The anti-inflammatory properties of AgNPs were assessed in vitro on HaCaT cells exposed to UVB radiation, in vivo on acute inflammation model and in humans on psoriasis lesions. In vitro, our results demonstrated the anti-inflammatory effects of functionalized AgNPs by the decrease of cytokines production induced by UVB irradiation. In vivo, the pre-administration of AgNPs reduced the edema and cytokines levels in the paw tissues, early after the induction of inflammation. The present study also demonstrated the possible use of synthesized AgNPs for the treatment of psoriasis lesions.
Sigma-Aldrich
Mouse IL-6 ELISA Kit, for serum, plasma and cell culture supernatant
Sigma-Aldrich
Mouse IL-6 ELISA Kit, for cell and tissue lysates
Sigma-Aldrich
Osmium tetroxide solution, 4 wt. % in H2O
Sigma-Aldrich
Osmium tetroxide solution, 2.5 wt. % in tert-butanol
Sigma-Aldrich
Sodium hydroxide-16O solution, 20 wt. % in H216O, 99.9 atom % 16O
Sigma-Aldrich
Osmium tetroxide solution, suitable for electron microscopy, 2% in H2O
Sigma-Aldrich
Osmium tetroxide solution, suitable for electron microscopy, 4% in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, 2%, solution
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, Molecular Biology, 0.5 M in H2O, DNase, RNase, NICKase and protease, none detected
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 50% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Glutaraldehyde solution, 50% in H2O, suitable for photographic applications
Sigma-Aldrich
Glutaraldehyde solution, Grade II, 25% in H2O
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 25% in H2O, specially purified for use as an electron microscopy fixative
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt dihydrate, reagent grade, 98.5-101.5% (titration)
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 8% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt dihydrate, Sigma Grade, suitable for plant cell culture, 98.5-101.5%
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 70% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt dihydrate, suitable for electrophoresis, Molecular Biology, 99.0-101.0% (titration)
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt dihydrate, BioUltra, 98.5-101.5%
Sigma-Aldrich
Osmium tetroxide, Sealed ampule.
Sigma-Aldrich
Glutaraldehyde solution, 25 wt. % in H2O
Sigma-Aldrich
Glutaraldehyde solution, 50 wt. % in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt dihydrate, ACS reagent, 99.0-101.0%
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Sigma-Aldrich
Glutaric dialdehyde solution, 50 wt. % in H2O, FCC
Supelco
Sodium hydroxide solution, 49-51% in water, eluent for IC
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt dihydrate, meets analytical specification of Ph. Eur., BP, USP, FCC, 99.0-101.0%
Sigma-Aldrich
Glutaraldehyde solution, technical, ~50% in H2O (5.6 M)
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt dihydrate, ≥97.0% (KT)