Merck
  • Stabilization of G-quadruplex DNA, inhibition of telomerase activity, and tumor cell apoptosis by organoplatinum(II) complexes with oxoisoaporphine.
Stabilization of G-quadruplex DNA, inhibition of telomerase activity, and tumor cell apoptosis by organoplatinum(II) complexes with oxoisoaporphine.
Journal of medicinal chemistry (2015-02-05)
Zhen-Feng Chen, Qi-Pin Qin, Jiao-Lan Qin, Yan-Cheng Liu, Ke-Bin Huang, Yu-Lan Li, Ting Meng, Guo-Hai Zhang, Yan Peng, Xu-Jian Luo, Hong Liang
ABSTRACT
Two G-quadruplex ligands [Pt(L(a))(DMSO)Cl] (Pt1) and [Pt(L(b))(DMSO)Cl] (Pt2) have been synthesized and fully characterized. The two complexes are more selective for SK-OV-3/DDP tumor cells versus normal cells (HL-7702). It was found that both Pt1 and Pt2 could be a telomerase inhibitor targeting G-quadruplex DNA. This is the first report demonstrating that telomeric, c-myc, and bcl-2 G-quadruplexes and caspase-3/9 preferred to bind with Pt2 rather than Pt1, which also can induce senescence and apoptosis. The different biological behavior of Pt1 and Pt2 may correlate with the presence of a 6-hydroxyl group in L(b). Importantly, Pt1 and Pt2 exhibited higher safety in vivo and more effective inhibitory effects on tumor growth in the HCT-8 and NCI-H460 xenograft mouse model, compared with cisplatin. Taken together, these mechanistic insights indicate that both Pt1 and Pt2 display low toxicity and could be novel anticancer drug candidates.
Sigma-Aldrich
Ethyl acetate, ≥99%, FCC, FG
Sigma-Aldrich
Ethyl acetate, anhydrous, 99.8%
Sigma-Aldrich
Ethyl acetate, ReagentPlus®, ≥99.8%
Sigma-Aldrich
Propidium iodide solution
Supelco
Ethyl acetate, analytical standard
Sigma-Aldrich
Ethyl acetate, natural, ≥99%, FCC, FG
Sigma-Aldrich
Ethyl acetate, suitable for HPLC, ≥99.8%
Sigma-Aldrich
Ethyl acetate, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Ethyl acetate, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%
Sigma-Aldrich
Ethyl acetate
Sigma-Aldrich
Ethyl acetate, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl acetate, biotech. grade, ≥99.8%
Sigma-Aldrich
Ethyl acetate, ACS reagent, ≥99.5%
Supelco
Ethyl Acetate, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Propidium iodide, ≥94% (HPLC)
Sigma-Aldrich
Thiazole Orange, Dye content ~90 %
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, 98%
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sigma-Aldrich
Zirconyl chloride octahydrate, reagent grade, 98%