Merck
  • Simultaneous quantification of purine and pyrimidine bases, nucleosides and their degradation products in bovine blood plasma by high performance liquid chromatography tandem mass spectrometry.
Simultaneous quantification of purine and pyrimidine bases, nucleosides and their degradation products in bovine blood plasma by high performance liquid chromatography tandem mass spectrometry.
Journal of chromatography. A (2014-07-16)
Charlotte Stentoft, Mogens Vestergaard, Peter Løvendahl, Niels Bastian Kristensen, Jon M Moorby, Søren Krogh Jensen
ABSTRACT
Improved nitrogen utilization in cattle is important in order to secure a sustainable cattle production. As purines and pyrimidines (PP) constitute an appreciable part of rumen nitrogen, an improved understanding of the absorption and intermediary metabolism of PP is essential. The present work describes the development and validation of a sensitive and specific method for simultaneous determination of 20 purines (adenine, guanine, guanosine, inosine, 2'-deoxyguanosine, 2'-deoxyinosine, xanthine, hypoxanthine), pyrimidines (cytosine, thymine, uracil, cytidine, uridine, thymidine, 2'-deoxyuridine), and their degradation products (uric acid, allantoin, β-alanine, β-ureidopropionic acid, β-aminoisobutyric acid) in blood plasma of dairy cows. The high performance liquid chromatography-based technique coupled to electrospray ionization tandem mass spectrometry (LC-MS/MS) was combined with individual matrix-matched calibration standards and stable isotopically labelled reference compounds. The quantitative analysis was preceded by a novel pre-treatment procedure consisting of ethanol precipitation, filtration, evaporation and reconstitution. Parameters for separation and detection during the LC-MS/MS analysis were investigated. It was confirmed that using a log-calibration model rather than a linear calibration model resulted in lower CV% and a lack of fit test demonstrated a satisfying linear regression. The method covers concentration ranges for each metabolite according to that in actual samples, e.g. guanine: 0.10-5.0 μmol/L, and allantoin: 120-500 μmol/L. The CV% for the chosen quantification ranges were below 25%. The method has good repeatability (CV%≤25%) and intermediate precision (CV%≤25%) and excellent recoveries (91-107%). All metabolites demonstrated good long-term stability and good stability within-runs (CV%≤10%). Different degrees of absolute matrix effects were observed in plasma, urine and milk. The determination of relative matrix effects revealed that the method was suitable for almost all examined PP metabolites in plasma drawn from an artery and the portal hepatic, hepatic and gastrosplenic veins and, with a few exceptions, also for other species such as chicken, pig, mink, human and rat.
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
2′-Deoxyinosine, ≥98%
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Adenine, European Pharmacopoeia (EP) Reference Standard
Allantoin, European Pharmacopoeia (EP) Reference Standard
Supelco
Allantoin, analytical standard
Supelco
Guanine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, Molecular Biology, 10 M in H2O
Sigma-Aldrich
Sodium hydroxide, pellets, semiconductor grade, 99.99% trace metals basis
Sigma-Aldrich
Sodium hydroxide, BioXtra, ≥98% (acidimetric), pellets (anhydrous)
Supelco
Sodium hydroxide solution, 49-51% in water, eluent for IC
Sigma-Aldrich
Sodium hydroxide solution, 50% in H2O
Sigma-Aldrich
Sodium hydroxide, reagent grade, 97%, flakes
Sigma-Aldrich
Sodium hydroxide, BioUltra, suitable for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
Sodium hydroxide, ACS reagent, ≥97.0%, pellets
Sigma-Aldrich
Sodium hydroxide, beads, 16-60 mesh, reagent grade, 97%
Sigma-Aldrich
Sodium hydroxide, reagent grade, ≥98%, pellets (anhydrous)
Sigma-Aldrich
Sodium hydroxide solution, 5.0 M
Sigma-Aldrich
Sodium hydroxide, reagent grade, 97%, powder
Sigma-Aldrich
Sodium hydroxide, puriss., meets analytical specification of Ph. Eur., BP, NF, E524, 98-100.5%, pellets
Sigma-Aldrich
Uridine, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Guanine, 98%
Sigma-Aldrich
Adenine, BioReagent, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Adenine, ≥99%
Sigma-Aldrich
Adenine, BioReagent, suitable for cell culture
Sigma-Aldrich
Acetic acid, glacial, ACS reagent, ≥99.7%
Sigma-Aldrich
Acetic acid, suitable for luminescence, BioUltra, ≥99.5% (GC)
Supelco
Acetic acid, analytical standard
Sigma-Aldrich
Acetic acid, glacial, ReagentPlus®, ≥99%