Merck
  • Francisella tularensis LVS surface and membrane proteins as targets of effective post-exposure immunization for tularemia.
Francisella tularensis LVS surface and membrane proteins as targets of effective post-exposure immunization for tularemia.
Journal of proteome research (2014-12-17)
Jeffrey C Chandler, Marjorie D Sutherland, Marisa R Harton, Claudia R Molins, Rebecca V Anderson, Darragh G Heaslip, Catharine M Bosio, John T Belisle
ABSTRACT
Francisella tularensis causes disease (tularemia) in a large number of mammals, including man. We previously demonstrated enhanced efficacy of conventional antibiotic therapy for tularemia by postexposure passive transfer of immune sera developed against a F. tularensis LVS membrane protein fraction (MPF). However, the protein composition of this immunogenic fraction was not defined. Proteomic approaches were applied to define the protein composition and identify the immunogens of MPF. MPF consisted of at least 299 proteins and 2-D Western blot analyses using sera from MPF-immunized and F. tularensis LVS-vaccinated mice coupled to liquid chromatography-tandem mass spectrometry identified 24 immunoreactive protein spots containing 45 proteins. A reverse vaccinology approach that applied labeling of F. tularensis LVS surface proteins and bioinformatics was used to reduce the complexity of potential target immunogens. Bioinformatics analyses of the immunoreactive proteins reduced the number of immunogen targets to 32. Direct surface labeling of F. tularensis LVS resulted in the identification of 31 surface proteins. However, only 13 of these were reactive with MPF and/or F. tularensis LVS immune sera. Collectively, this use of orthogonal proteomic approaches reduced the complexity of potential immunogens in MPF by 96% and allowed for prioritization of target immunogens for antibody-based immunotherapies against tularemia.
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Supelco
Glycerol, analytical standard
Sodium laurilsulfate, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Supelco
Urea, analytical standard
Sigma-Aldrich
Ammonium bicarbonate, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Glycerol, ReagentPlus®, ≥99.0% (GC)
Sigma-Aldrich
Glycerol, ACS reagent, ≥99.5%
Sigma-Aldrich
Formic acid, reagent grade, ≥95%
Sigma-Aldrich
Formic acid, ACS reagent, ≥88%
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
USP
Residual Solvent Class 2 - Acetonitrile, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Glycerol, FCC, FG
Sigma-Aldrich
Bromophenol Blue, ACS reagent
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
Ammonium bicarbonate, BioUltra, ≥99.5% (T)
Sigma-Aldrich
Glycerol, ≥99.5%
Sigma-Aldrich
Glycerol, Molecular Biology, ≥99.0%
Sigma-Aldrich
Glycerol, BioXtra, ≥99% (GC)
Sigma-Aldrich
Urea, meets USP testing specifications
Sigma-Aldrich
Urea, suitable for electrophoresis
Sigma-Aldrich
Urea, BioXtra, pH 7.5-9.5 (20 °C, 5 M in H2O)
Sigma-Aldrich
Urea, powder, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Urea, ACS reagent, 99.0-100.5%
Sigma-Aldrich
Bromophenol Blue, titration: suitable
Sigma-Aldrich
Glycerin, meets USP testing specifications
Sigma-Aldrich
Glycerol, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)