Merck
  • Avian prostatic acid phosphatase: estrogen regulation in the oviduct and epithelial cell-derived ovarian carcinomas.
Avian prostatic acid phosphatase: estrogen regulation in the oviduct and epithelial cell-derived ovarian carcinomas.
Biology of reproduction (2014-05-16)
Hyocheol Bae, Whasun Lim, Seung-Min Bae, Fuller W Bazer, Youngsok Choi, Gwonhwa Song
ABSTRACT
Prostatic acid phosphatase (ACPP) is a glycoprotein that is mainly synthesized and secreted by glandular epithelial cells (GE) of the prostate, and it is well known as a biomarker for prostate cancer. Although ACPP was used as prognostic/diagnostic indicator and studied to elucidate regulatory mechanism(s) during several decades in humans, its role is not clearly understood. Gene profiling data using a chicken DNA microarray revealed that ACPP increased significantly during remodeling and recrudescence of the oviduct in response to estrogen. Thus, in this study, we investigated the expression and hormonal regulation of ACPP gene in the reproductive tracts of chickens. ACPP was specifically detected in the luminal cells (LE) and GE of chicken oviduct, and diethylstilbestrol (a synthetic nonsteroidal estrogen) stimulated its expression during development of the oviduct. In addition, ACPP mRNA and protein were localized to LE and GE during the regeneration phase of the oviduct of laying hens during induced molting. Furthermore, ACPP mRNA and protein were abundant in GE of ovarian carcinoma, but not in normal ovaries. Moreover, strong expression of ACPP protein was detected in epithelial cells of cancerous ovaries from women. Collectively, results of the present study are the first to show that ACPP is a novel estrogen-stimulated gene in the oviductal epithelial cells of the chicken and that its expression increases significantly in epithelial cells of ovarian carcinoma, which indicates that it may be a candidate biomarker for diagnosis of epithelia-derived ovarian cancer in women.
Product Description
Supelco
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Supelco
Digoxigenin, analytical standard
USP
Dehydrated Alcohol, United States Pharmacopeia (USP) Reference Standard
Digoxigenin, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Ethyl alcohol, Pure 190 proof, for molecular biology
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Ethanol, BioUltra, Molecular Biology, ≥99.8%, (absolute alcohol, without additive, A15 o1)
Sigma-Aldrich
Ethanol, tested according to Ph. Eur.
Sigma-Aldrich
Ethanol
Supelco
Ethanol, standard for GC
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Ethyl alcohol, Pure 200 proof, Molecular Biology
Sigma-Aldrich
Ethanol, purum, fine spirit, denaturated with 4.8% methanol, F25 METHYL1, ~96% (based on denaturant-free substance)
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications