Merck
  • Electrospun scaffolds for multiple tissues regeneration in vivo through topography dependent induction of lineage specific differentiation.
Electrospun scaffolds for multiple tissues regeneration in vivo through topography dependent induction of lineage specific differentiation.
Biomaterials (2015-01-27)
Zi Yin, Xiao Chen, Hai-Xin Song, Jia-Jie Hu, Qiao-Mei Tang, Ting Zhu, Wei-Liang Shen, Jia-Lin Chen, Huanhuan Liu, Boon Chin Heng, Hong-Wei Ouyang
ABSTRACT
Physical topographic cues from various substrata have been shown to exert profound effects on the growth and differentiation of stem cells due to their niche-mimicking features. However, the biological function of different topographic materials utilized as bio-scaffolds in vivo have not been rigorously characterized. This study investigated the divergent differentiation pathways of mesenchymal stem cells (MSCs) and neo-tissue formation trigged by aligned and randomly-oriented fibrous scaffolds, both in vitro and in vivo. The aligned group was observed to form more mature tendon-like tissue in the Achilles tendon injury model, as evidenced by histological scoring and collagen I immunohistochemical staining data. In contrast, the randomly-oriented group exhibited much chondrogenesis and subsequent bone tissue formation through ossification. Additionally, X-ray imaging and osteocalcin immunohistochemical staining also demonstrated that osteogenesis in vivo is driven by randomly oriented topography. Furthermore, MSCs on the aligned substrate exhibited tenocyte-like morphology and enhanced tenogenic differentiation compared to cells grown on randomly-oriented scaffold. qRT-PCR analysis of osteogenic marker genes and alkaline phosphatase (ALP) staining demonstrated that MSCs cultured on randomly-oriented fiber scaffolds displayed enhanced osteogenic differentiation compared with cells cultured on aligned fiber scaffolds. Finally, it was demonstrated that cytoskeletal tension release abrogated the divergent differentiation pathways on different substrate topography. Collectively, these findings illustrate the relationship between topographic cues of the scaffold and their inductive role in tissue regeneration; thus providing an insight into future development of smart functionalized bio-scaffold design and its application in tissue engineering.
Sigma-Aldrich
L-Ascorbic acid, meets USP testing specifications
Sigma-Aldrich
L-Ascorbic acid, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
L-Ascorbic acid, reagent grade
Sigma-Aldrich
L-Ascorbic acid, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
L-Ascorbic acid, reagent grade, crystalline
Sigma-Aldrich
L-Ascorbic acid, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
L-Ascorbic acid, 99%
Sigma-Aldrich
L-Ascorbic acid, ACS reagent, ≥99%
Sigma-Aldrich
L-Ascorbic acid, BioUltra, ≥99.5% (RT)
Sigma-Aldrich
L-Ascorbic acid, tested according to Ph. Eur.
Sigma-Aldrich
L-Ascorbic acid, FCC, FG
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ≥99.0% (RT)
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)
Supelco
L-Ascorbic acid, analytical standard
Supelco
L-Ascorbic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Edelfosine, ≥95% (HPLC)
Dexamethasone for peak identification, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Dexamethasone, powder, BioReagent, suitable for cell culture, ≥97%
Sigma-Aldrich
Dexamethasone, ≥98% (HPLC), powder
Sigma-Aldrich
Dexamethasone, powder, γ-irradiated, BioXtra, suitable for cell culture, ≥80% (HPLC)
Sigma-Aldrich
Dexamethasone, meets USP testing specifications
Sigma-Aldrich
Dexamethasone, tested according to Ph. Eur.
Supelco
Dexamethasone, VETRANAL®, analytical standard
Supelco
Ascorbic Acid, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Ascorbic acid, United States Pharmacopeia (USP) Reference Standard
Dexamethasone, European Pharmacopoeia (EP) Reference Standard
Dexamethasone, British Pharmacopoeia (BP) Assay Standard
Supelco
Dexamethasone, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Dexamethasone, United States Pharmacopeia (USP) Reference Standard