Merck
  • Self-nanoemulsifying drug delivery systems for oral insulin delivery: in vitro and in vivo evaluations of enteric coating and drug loading.
Self-nanoemulsifying drug delivery systems for oral insulin delivery: in vitro and in vivo evaluations of enteric coating and drug loading.
International journal of pharmaceutics (2014-12-03)
Ping Li, Angel Tan, Clive A Prestidge, Hanne Mørck Nielsen, Anette Müllertz
ABSTRACT
This study aims at evaluating the combination of self-nanoemulsifying drug delivery systems (SNEDDS) and enteric-coated capsules as a potential delivery strategy for oral delivery of insulin. The SNEDDS preconcentrates, loaded with insulin-phospholipid complex at different levels (0, 2.5 and 10% w/w), were readily dispersed in water to form nanoemulsions of 35 nm and vesicles of 300 nm. The association efficiency of non-complexed insulin in the dispersed SNEDDS was 18.6%, and was increased to 73.1% for insulin-phospholipid complex (at 10% loading level). The morphology of the dispersed SNEDDS changed from nanoemulsion droplets to vesicular structures with increasing complex loading levels. A pH-dependent insulin release profile was observed for SNEDDS filled into capsules coated with the enteric polymer, Eudragit(®) L100. Using a Caco-2 cell model, it was observed that the transport of insulin was enhanced by factors of 7.7- and 9.3- for SNEDDS loaded with 2.5 and 10% complex, respectively. In healthy fasted rats, administration of SNEDDS (10% complex) filled in enteric-coated capsules produced a 2.7-fold and 3.4-fold enhancement in the relative bioavailability and glucose reduction, respectively. This study shows the effectiveness of combining SNEDDS (loaded with insulin-phospholipid complex) with enteric-coated capsules for enhancing the oral absorption and efficacy of insulin.
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
1-Decanoyl-rac-glycerol, ≥99%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
HEPES buffer solution, 1 M in H2O
Sigma-Aldrich
Ethyl alcohol, Pure 200 proof, Molecular Biology
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
Sigma-Aldrich
Ethanol
Sigma-Aldrich
Ethanol, BioUltra, Molecular Biology, ≥99.8%, (absolute alcohol, without additive, A15 o1)
Sigma-Aldrich
Ethanol, purum, fine spirit, denaturated with 4.8% methanol, F25 METHYL1, ~96% (based on denaturant-free substance)
Supelco
Ethanol, standard for GC
Sigma-Aldrich
Ethanol, tested according to Ph. Eur.
USP
Dehydrated Alcohol, United States Pharmacopeia (USP) Reference Standard
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Supelco
HEPES, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
HEPES, anhydrous, free-flowing, Redi-Dri, ≥99.5%
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
1-(2-Hydroxyethyl)piperazine, 98%
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioUltra, Molecular Biology, ≥99.5% (T)
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
Ethyl alcohol, Pure 190 proof, for molecular biology
Supelco
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material