Merck
  • Accurate analysis of ginkgolides and their hydrolyzed metabolites by analytical supercritical fluid chromatography hybrid tandem mass spectrometry.
Accurate analysis of ginkgolides and their hydrolyzed metabolites by analytical supercritical fluid chromatography hybrid tandem mass spectrometry.
Journal of chromatography. A (2015-03-03)
Xin-Guang Liu, Lian-Wen Qi, Zhi-Ying Fan, Xin Dong, Ru-Zhou Guo, Feng-Chang Lou, Salvatore Fanali, Ping Li, Hua Yang
ABSTRACT
Hydrolysis plays an important role in the metabolic transformations of lactones. Analysis of lactones and their hydrolyzed metabolites in biological samples is challenging, because unexpected hydrolysis or reversed dehydration may occur depending on the test environments. In this work, a supercritical fluid chromatography hybrid triple-quadrupole mass spectrometry (SFC-QQQ MS) method has been proposed for simultaneous analysis of ginkgolides and hydrolyzed metabolites. The SFC utilizes carbon dioxide as the mobile phase, avoiding hydrolysis of ginkgolides that always happens during reversed phase liquid chromatographic detection. Compared with normal phase liquid chromatography, the SFC provides good resolutions especially for ginkgolide with similar structures. This SFC-QQQ MS method was validated in linearity, precision, accuracy, and stability for ginkgolides and metabolites. Then this method was successfully applied to pharmacokinetic study of 3 ginkgolides and their 6 hydrolyzed metabolites after intravenous administration of total ginkgolide extract. Ginkgolides and metabolites showed different clear rates and excluded in 2-4h. The developed SFC-QQQ MS method allows accurate determination of ginkgolides and metabolites with high resolutions, and can be extended to analysis of other water-unstable compounds.
Product Description
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
USP
Glycerin, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Sodium hydroxide, ultra dry, powder or crystals, 99.99% trace metals basis
Sigma-Aldrich
Sodium hydroxide solution, 5.0 M
Sigma-Aldrich
Sodium hydroxide, ACS reagent, ≥97.0%, pellets
Sigma-Aldrich
Sodium hydroxide, reagent grade, 97%, powder
Sigma-Aldrich
Sodium hydroxide solution, 50% in H2O
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Sodium hydroxide, reagent grade, ≥98%, pellets (anhydrous)
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Sodium hydroxide, reagent grade, 97%, flakes
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Ethyl alcohol, Pure 190 proof, for molecular biology
Sigma-Aldrich
Sodium hydroxide, BioUltra, suitable for luminescence, ≥98.0% (T), pellets
Supelco
Sodium hydroxide solution, 49-51% in water, eluent for IC
Supelco
Sodium hydroxide concentrate, 0.1 M NaOH in water (0.1N), Eluent concentrate for IC
Supelco
Methanol, analytical standard
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, Molecular Biology, 10 M in H2O
Supelco
Glycerin, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
USP
Dehydrated Alcohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Sodium hydroxide, beads, 16-60 mesh, reagent grade, 97%
Sigma-Aldrich
Sodium hydroxide, pellets, semiconductor grade, 99.99% trace metals basis
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%