Merck
Down-expression pattern of Ku70 and p53 coexisted in colorectal cancer.
Medical oncology (Northwood, London, England) (2015-03-04)
Yuanfang Lu, Jingyan Gao, Yuanming Lu
ABSTRACT
To address the relationship of altered expression of double-strand break repair proteins Ku70 and p53 in clinical colorectal cancer (CRC), we examined the expression pattern of Ku70 and p53 by using fluorescent immunohistochemistry and real-time PCR assays in CRC and pericancerous samples from 152 Chinese patients. The results showed that down-expression pattern of both Ku70 and p53 coexisted in the CRC samples with significant correlating rate (R (2) = 0.9103; P < 0.001), and the down-expression of Ku70 and p53 was significantly associated with the advanced tumor node metastasis stage (Ku70: HR 3.453 in recurrence and 4.182 in survival, P < 0.001; P53: HR 3.114 in recurrence and 4.113 in survival, P < 0.001). The down-regulated Ku70 and p53 were associated with poor disease-free survival. Loss of Ku70 and p53 expression might serve as a biomarker of poor prognosis in CRC patients.
Sigma-Aldrich
Formaldehyde-12C solution, 20% in H2O, 99.9 atom % 12C
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, suitable for protein labeling, ≥90% (HPLC), powder
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, ≥97.5% (HPLC)
Sigma-Aldrich
Formaldehyde solution, tested according to Ph. Eur.
Sigma-Aldrich
Formaldehyde solution, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
Sigma-Aldrich
Formaldehyde solution, Molecular Biology, BioReagent, ≥36.0% in H2O (T)
Sigma-Aldrich
Formaldehyde solution, meets analytical specification of USP, ≥34.5 wt. %
Sigma-Aldrich
Formaldehyde solution, Molecular Biology, 36.5-38% in H2O
Supelco
Formaldehyde solution, stabilized with methanol, ~37 wt. % in H2O, certified reference material
SAFC
Formaldehyde solution, contains 10-15% methanol as stabilizer, 37 wt. % in H2O
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, ≥90% (HPLC)
Sigma-Aldrich
Hematoxylin
Sigma-Aldrich
Hematoxylin, certified by the BSC
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, BioReagent, suitable for fluorescence, mixture of 2 components, ≥90% (HPLC)
Sigma-Aldrich
Xylenes, reagent grade
Sigma-Aldrich
Xylenes, histological grade
Sigma-Aldrich
Xylenes, ACS reagent, ≥98.5% xylenes + ethylbenzene basis