Merck
  • Next-generation steroidogenesis inhibitors, dutasteride and abiraterone, attenuate but still do not eliminate androgen biosynthesis in 22RV1 cells in vitro.
Next-generation steroidogenesis inhibitors, dutasteride and abiraterone, attenuate but still do not eliminate androgen biosynthesis in 22RV1 cells in vitro.
The Journal of steroid biochemistry and molecular biology (2014-09-10)
Steven Pham, Subrata Deb, Dong Sheng Ming, Hans Adomat, Elham Hosseini-Beheshti, Amina Zoubeidi, Martin Gleave, Emma S Tomlinson Guns
ABSTRACT
Castration resistant prostate cancer (CRPC) is often lethal and inevitably develops after androgen ablation therapy. However, in the majority of cases it remains androgen dependent. CRPC tumors have the ability to synthesize their own androgens from cholesterol by engaging in de novo steroidogenesis. We investigated the potential of 22RV1 prostate cancer cells to convert the supplemented steroid precursors within this pathway under the effects of current clinical steroidogenesis inhibitors such as abiraterone and dutasteride, either alone or in combination. Under steroid starved conditions, enzymes responsible for de novo steroidogenesis were upregulated. Testosterone and dihydrotestosterone (DHT) were formed by using both dehydroepiandrosterone (DHEA) and progesterone as substrates. Formation of testosterone and DHT was higher following incubation with DHEA compared to progesterone. Progesterone decreased the mRNA expression of enzymes responsible for steroidogenesis. Abiraterone treatment decreased testosterone production but increased several precursor steroids in both classical and backdoor pathways in the presence of progesterone. In contrast, the DHT levels were elevated following treatment with abiraterone when progesterone was absent. Dutasteride decreased the formation of testosterone, DHT and precursor steroids in the backdoor pathway but increased steroid precursors in the classical steroidogenesis pathway. The combination of abiraterone and dutasteride decreased testosterone and DHT in the presence of progesterone but increased DHT in the absence of progesterone. Abiraterone inhibited androgen receptor (AR) activation but not to the same extent as MDV3100. However, abiraterone and dutasteride treatment, either alone or in combination, were more effective in decreasing prostate specific antigen secretion into the media than MDV3100. Thus, while interventions with these drugs alone or in combination fail to completely inhibit steroidogenesis in the 22RV1 cells, the combined inhibition of androgen production and blockade of AR can exceed the effect of MDV3100. Further characterization of bypass mechanisms that may develop as a response to these inhibitors is necessary to achieve optimal suppression of testosterone and DHT synthesis as a part of therapeutic regimens for the treatment of CRPC.
Product Description
Supelco
Acetonitrile(Neat), Pharmaceutical Secondary Standard; Certified Reference Material
Pricing and availability is not currently available.
Supelco
Triethylamine, analytical standard
Pricing and availability is not currently available.
Sigma-Aldrich
Testosterone, purum, ≥99.0% (HPLC)
Pricing and availability is not currently available.
Sigma-Aldrich
Triethylamine, puriss. p.a., ≥99.5% (GC)
Pricing and availability is not currently available.
Sigma-Aldrich
Triethylamine, ≥99.5%
Pricing and availability is not currently available.
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Pricing and availability is not currently available.
Sigma-Aldrich
Triethylamine, for amino acid analysis, ≥99.5% (GC)
Pricing and availability is not currently available.
Sigma-Aldrich
Triethylamine, BioUltra, ≥99.5% (GC)
Pricing and availability is not currently available.
Sigma-Aldrich
trans-Dehydroandrosterone, ≥99%
Pricing and availability is not currently available.
Sigma-Aldrich
Triethylamine, ≥99%
Pricing and availability is not currently available.
Sigma-Aldrich
Triethylamine, ≥99.5%
Pricing and availability is not currently available.
Supelco
Methanol, analytical standard
Pricing and availability is not currently available.
Supelco
Testosterone, VETRANAL®, analytical standard
Pricing and availability is not currently available.
Supelco
Testosterone solution, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®
Pricing and availability is not currently available.
Testosterone, European Pharmacopoeia (EP) Reference Standard
Pricing and availability is not currently available.
Supelco
Androsterone, VETRANAL®, analytical standard
Pricing and availability is not currently available.
Supelco
Residual Solvent - Acetonitrile(solution in DMSO), Pharmaceutical Secondary Standard; Certified Reference Material
Pricing and availability is not currently available.
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Pricing and availability is not currently available.
Sigma-Aldrich
Methanol, Absolute - Acetone free
Pricing and availability is not currently available.
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Pricing and availability is not currently available.
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Pricing and availability is not currently available.
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Pricing and availability is not currently available.
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Pricing and availability is not currently available.
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Pricing and availability is not currently available.
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Pricing and availability is not currently available.
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Pricing and availability is not currently available.
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Pricing and availability is not currently available.
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Pricing and availability is not currently available.
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Pricing and availability is not currently available.
Sigma-Aldrich
Triethylamine, ampule, ≥99.5% (GC)
Pricing and availability is not currently available.