Merck
  • Semicarbazide-sensitive amine oxidase (SSAO) inhibition ameliorates kidney fibrosis in a unilateral ureteral obstruction murine model.
Semicarbazide-sensitive amine oxidase (SSAO) inhibition ameliorates kidney fibrosis in a unilateral ureteral obstruction murine model.
American journal of physiology. Renal physiology (2014-08-22)
May Wong, Sonia Saad, Jie Zhang, Simon Gross, Wolfgang Jarolimek, Heidi Schilter, Jason A Chen, Anthony J Gill, Carol A Pollock, Muh Geot Wong
ABSTRACT
Semicarbazide-sensitive amine oxidase (SSAO) is an enzyme known for its dual function in mediating inflammation and reactive oxygen species production. However, the role of SSAO inhibitors in limiting kidney fibrosis is unclear. We aimed to determine the effectiveness of a SSAO inhibitor (SSAOi; PXS4728A) as an antifibrotic agent using a 7-day unilateral ureteric obstruction (UUO) model of acute kidney fibrosis in 6- to 8-wk-old mice. The experimental groups were 1) Sham operated; 2) UUO; 3) UUO+SSAOi (2 mg/kg); 4) UUO+telmisartan, an angiotensin receptor blocker (3 mg/kg); and 5) UUO+SSAOi+telmisartan. Kidney tissue was analyzed for histological evidence of tubulointerstitial fibrosis, nitrotyrosine staining, and mRNA expression of markers associated with fibrosis and inflammation. Kidney SSAO activity was determined by radiometric [(14)C]benzylamine methodology. Our results show that SSAOi effectively suppresses UUO-mediated SSAO activity. Extracellular matrix markers, namely, fibronectin, collagen IV protein, and nitrotyrosine staining, were lower in UUO+SSAOi mice compared with untreated UUO mice. This was consistent with the attenuated mRNA expression of collagen IV and fibronectin. SSAOi effectively inhibited transforming growth factor-β1 (TGF-β1) and monocyte chemoattractant protein-1 (MCP-1) expression to a similar extent to that observed with telmisartan. Individually, SSAOi and telmisartan induced a reduction in interstitial leukocyte and macrophage accumulation. However, the combination of SSAOi and telmisartan was more effective at reducing inflammatory cell infiltration. These results demonstrate that SSAO inhibition significantly suppresses profibrotic and proinflammatory cytokine secretion, reduces oxidative stress, and limits inflammatory cell accumulation and extracellular matrix expression in an acute model of renal fibrosis.
Sigma-Aldrich
Toluene, ACS reagent, ≥99.5%
Sigma-Aldrich
Toluene, ACS reagent, ≥99.5%
Sigma-Aldrich
Toluene, ACS reagent, ≥99.5%
Sigma-Aldrich
Toluene, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%
Sigma-Aldrich
Toluene, anhydrous, 99.8%
Supelco
Toluene, analytical standard
Supelco
Toluene, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Residual Solvent - Toluene, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Hydrogen peroxide solution, 30 % (w/w) in H2O, contains stabilizer
Supelco
Hydrogen peroxide solution, ≥30%, for trace analysis
Millipore
Hydrogen peroxide solution, 3%, suitable for microbiology
Sigma-Aldrich
Hydrogen peroxide solution, 34.5-36.5%
Sigma-Aldrich
Hydrogen peroxide solution, tested according to Ph. Eur.
Supelco
Hydrogen peroxide solution, 30 % (w/w), for ultratrace analysis
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 30 wt. % in H2O, meets USP testing specifications
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 30 wt. % in H2O, ACS reagent
Sigma-Aldrich
Hydrogen peroxide solution, 50 wt. % in H2O, stabilized