Merck
  • Separation and detection of plasmalogen in marine invertebrates by high-performance liquid chromatography with evaporative light-scattering detection.
Separation and detection of plasmalogen in marine invertebrates by high-performance liquid chromatography with evaporative light-scattering detection.
Lipids (2014-11-12)
Shinji Yamashita, Akihiro Abe, Kiyotaka Nakagawa, Mikio Kinoshita, Teruo Miyazawa
ABSTRACT
We have developed a new method for determining ethanolamine plasmalogen contents in marine invertebrates. This quantification method involves derivatization of ethanolamine glycerophospholipid (EtnGpl) subclasses, alkenylacyl (plasmalogen), diacyl, and alkylacyl subclasses, by enzyme treatment and acetylation, followed by separation and detection by high-performance liquid chromatography (HPLC) with evaporative light-scattering detection (ELSD). This method enabled complete separation of the subclasses, and the limit of detection for plasmalogen was 200 ng (260 pmol). The peak area of plasmalogen by ELSD was unaffected by the degree of unsaturated fatty acids in EtnGpl, in contrast to ultraviolet (UV) detection. Thus, this method enables accurate determination of plasmalogen contents in various species containing marine products possessing abundant polyunsaturated fatty acids (PUFA). The method developed here was applied to marine invertebrates available in Japan. The examined marine invertebrates showed a wide range of plasmalogen contents ranging from 19 to 504 μmol/100 g wet wt. The plasmalogen levels in samples except those of class Cephalopoda and Crustacea were more than 60 mol% of EtnGpl.
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Chloroform, anhydrous, ≥99%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%
Supelco
Chloroform, analytical standard
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Chloroform, ≥99%, PCR Reagent, contains amylenes as stabilizer
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Chloroform, contains 100-200 ppm amylenes as stabilizer, ≥99.5%
Supelco
Methanol, analytical standard
Sigma-Aldrich
Chloroform, contains ethanol as stabilizer, meets analytical specification of BP, 99-99.4% (GC)
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Chloroform, biotech. grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
O-Phosphorylethanolamine
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Chloroform, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains amylenes as stabilizer
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Chloroform, contains ethanol as stabilizer, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Chloroform, ACS spectrophotometric grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, contains amylenes as stabilizer, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Chloroform, suitable for HPLC, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Supelco
Chloroform, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Chloroform, ReagentPlus®, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer