Merck
  • Protective effect of resveratrol on sodium fluoride-induced oxidative stress, hepatotoxicity and neurotoxicity in rats.
Protective effect of resveratrol on sodium fluoride-induced oxidative stress, hepatotoxicity and neurotoxicity in rats.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association (2014-05-27)
Nurgul Atmaca, Hasan Tarik Atmaca, Ayse Kanici, Tugce Anteplioglu
ABSTRACT
Protective effect of resveratrol on sodium fluoride-induced oxidative stress, hepatotoxicity and neurotoxicity were studied in rats. A total of 28 Wistar albino male rats were used. Four study groups were randomly formed with seven animals in each. The groups were treated for 21days with distilled water (control group), with water containing 100ppm fluoride (fluoride group), with resveratrol (12.5mg/kg i.p., resveratrol group), or with 100ppm fluoride+12.5mg/kg resveratrol i.p. (fluoride+resveratrol group). At the end of the trial, blood samples were collected by cardiac puncture and tissue samples were taken simultaneously. The total antioxidant and oxidant status in plasma and tissues as well as plasma 8-hydroxydeoxyguanosine levels were measured. Histopathological analyses of rat liver and brain tissues were performed in all groups to identify any changes. In the fluoride group, the total oxidant levels increased in plasma, liver and brain and total antioxidant levels decreased, as did the plasma 8-hydroxy-deoxyguanosine levels. These changes were prevented by co-administration of resveratrol. In addition, fluoride-associated severe histopathological changes in brain and liver tissues were not observed in the fluoride+resveratrol group. Consequently, these data suggested that resveratrol had beneficial effects in alleviating fluoride-induced oxidative stress.
Sigma-Aldrich
Sodium Fluoride Solution
Sigma-Aldrich
Sodium fluoride, 99.99% trace metals basis
Sigma-Aldrich
Sodium fluoride, anhydrous, powder, 99.99% trace metals basis
Sigma-Aldrich
Sodium fluoride, BioReagent, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Sodium fluoride, BioXtra, ≥99%
Sigma-Aldrich
Sodium fluoride 0.5 M solution
Supelco
Sodium Fluoride, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Fluoride ion solution for ISE, 0.1 M F-, for ion-selective electrodes
Sigma-Aldrich
Sodium fluoride, ACS reagent, ≥99%
Sigma-Aldrich
Sodium fluoride, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium fluoride, puriss., meets analytical specification of Ph. Eur., BP, USP, 98.5-100.5% (calc. to the dried substance)