Merck
  • Design, synthesis and biological evaluation of 2-aminopyrimidinones and their 6-aza-analogs as a new class of CK2 inhibitors.
Design, synthesis and biological evaluation of 2-aminopyrimidinones and their 6-aza-analogs as a new class of CK2 inhibitors.
Journal of enzyme inhibition and medicinal chemistry (2013-10-05)
Maksym O Chekanov, Olga V Ostrynska, Sergii S Tarnavskyi, Anatoliy R Synyugin, Nadiia V Briukhovetska, Volodymyr G Bdzhola, Alexander E Pashenko, Andrey A Fokin, Sergiy M Yarmoluk
ABSTRACT
In order to find the new potent CK2 inhibitors the 60 derivatives of 2-aminopyrimidinone and their 6-aza-substituted analogs were synthesized and tested in vitro. Among them, the most efficient inhibitor 2-hydroxy-5-[4-(4-methoxyphehyl)-6-oxo-1,6-dihydropyrimidin-2-ylamino] benzoic acid was identified (IC50 = 1.1 μM). The structure--activity relationship study of newly synthesized derivatives was carried out and their binding mode with adenosine triphosphate-acceptor site of CK2 was proposed.
Sigma-Aldrich
Magnesium chloride solution, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Magnesium chloride solution, 0.1 M
Sigma-Aldrich
Magnesium chloride solution, PCR Reagent, 25 mM MgCI2 solution for PCR
Sigma-Aldrich
Magnesium chloride solution, BioUltra, Molecular Biology, ~0.025 M in H2O
Sigma-Aldrich
Magnesium chloride solution, BioUltra, Molecular Biology, 2 M in H2O
Sigma-Aldrich
Magnesium chloride solution, Molecular Biology, 1.00 M±0.01 M
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.9% trace metals basis
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Magnesium chloride, suitable for insect cell culture, BioReagent, ≥97.0%
Sigma-Aldrich
Magnesium chloride, ≥98%
Sigma-Aldrich
Magnesium chloride, powder, <200 μm