Merck
  • Effects of formulation and process factors on the crystal structure of freeze-dried Myo-inositol.
Effects of formulation and process factors on the crystal structure of freeze-dried Myo-inositol.
Journal of pharmaceutical sciences (2014-06-12)
Ken-Ichi Izutsu, Chikako Yomota, Haruhiro Okuda, Toru Kawanishi, Takuya Yamaki, Ryohei Ohdate, Zhaokun Yu, Etsuo Yonemochi, Katsuhide Terada
ABSTRACT
The objective of this study was to elucidate effects of formulation and process variables on the physical forms of freeze-dried myo-inositol. Physical properties of myo-inositol in frozen solutions, freeze-dried solids, and cooled heat-melt solids were characterized by powder X-ray diffraction (PXRD), thermal analysis (differential scanning calorimetry [DSC] and thermogravimetric), and simultaneous PXRD-DSC analysis. Cooling of heat-melt myo-inositol produced two forms of metastable anhydrate crystals that change to stable form (melting point 225 °C-228 °C) with transition exotherms at around 123 °C and 181 °C, respectively. Freeze-drying of single-solute aqueous myo-inositol solutions after rapid cooling induced crystallization of myo-inositol as metastable anhydrate (transition at 80 °C-125 °C) during secondary drying segment. Contrarily, postfreeze heat treatment (i.e., annealing) induced crystallization of myo-inositol dihydrate. Removal of the crystallization water during the secondary drying produced the stable-form myo-inositol anhydrate crystal. Shelf-ramp slow cooling of myo-inositol solutions resulted in the stable and metastable anhydrous crystal solids depending on the solute concentrations and the solution volumes. Colyophilization with phosphate buffer retained myo-inositol in the amorphous state. Crystallization in different process segments varies crystal form of freeze-dried myo-inositol solids.
Sigma-Aldrich
Sodium phosphate dibasic, Molecular Biology, ≥98.5% (titration)
Sigma-Aldrich
Sodium phosphate dibasic, 99.95% trace metals basis
Sigma-Aldrich
Sodium phosphate dibasic, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.0%, free-flowing, Redi-Dri
Sigma-Aldrich
Sodium phosphate dibasic, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Sodium phosphate dibasic, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium phosphate dibasic, purum p.a., anhydrous, ≥98.0% (T)
Sigma-Aldrich
Sodium phosphate dibasic, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E 339, anhydrous, 98-100.5% (calc. to the dried substance)
Sigma-Aldrich
Sodium phosphate dibasic, BioUltra, Molecular Biology, ≥99.5% (T)
Sigma-Aldrich
Sodium phosphate dibasic, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium phosphate dibasic, puriss. p.a., ACS reagent, anhydrous, ≥99.0% (T)
Sigma-Aldrich
Sodium phosphate dibasic, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.0%
Sigma-Aldrich
Sodium phosphate dibasic solution, BioUltra, 0.5 M in H2O
Millipore
myo-Inositol, ≥99.0%, suitable for microbiology
Sigma-Aldrich
myo-Inositol, ≥99%
Sigma-Aldrich
myo-Inositol, ≥99% (GC), BioReagent
Supelco
Phosphate Standard for IC, TraceCERT®, 1000 mg/L phosphate in water (nominal concentration)
myo-Inositol, European Pharmacopoeia (EP) Reference Standard