Merck
  • PP6 controls T cell development and homeostasis by negatively regulating distal TCR signaling.
PP6 controls T cell development and homeostasis by negatively regulating distal TCR signaling.
Journal of immunology (Baltimore, Md. : 1950) (2015-01-23)
Jian Ye, Hao Shi, Ye Shen, Chao Peng, Yan Liu, Chenyu Li, Kejing Deng, Jianguo Geng, Tian Xu, Yuan Zhuang, Biao Zheng, Wufan Tao
ABSTRACT
T cell development and homeostasis are both regulated by TCR signals. Protein phosphorylation and dephosphorylation, which are catalyzed by protein kinases and phosphatases, respectively, serve as important switches controlling multiple downstream pathways triggered by TCR recognition of Ags. It has been well documented that protein tyrosine phosphatases are involved in negative regulation of proximal TCR signaling. However, how TCR signals are terminated or attenuated in the distal TCR signaling pathways is largely unknown. We investigated the function of Ser/Thr protein phosphatase (PP) 6 in TCR signaling. T cell lineage-specific ablation of PP6 in mice resulted in enhanced thymic positive and negative selection, and preferential expansion of fetal-derived, IL-17-producing Vγ6Vδ1(+) T cells. Both PP6-deficient peripheral CD4(+) helper and CD8(+) cytolytic cells could not maintain a naive state and became fast-proliferating and short-lived effector cells. PP6 deficiency led to profound hyperactivation of multiple distal TCR signaling molecules, including MAPKs, AKT, and NF-κB. Our studies demonstrate that PP6 acts as a critical negative regulator, not only controlling both αβ and γδ lineage development, but also maintaining naive T cell homeostasis by preventing their premature activation before Ag stimulation.
Sigma-Aldrich
Dihydroethidium, ≥95%
Sigma-Aldrich
5(6)-Carboxyfluorescein diacetate N-succinimidyl ester, BioReagent, suitable for fluorescence, ≥90% (HPLC)
Sigma-Aldrich
Dihydroethidium, BioReagent, suitable for fluorescence, ≥95% (HPCE)
Sigma-Aldrich
5-Carboxy-fluorescein diacetate N-succinimidyl ester, suitable for fluorescence, ≥95.0% (HPLC)
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥98.5% (GC)
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥99.0% (T)
Sigma-Aldrich
Anti-PP6C Antibody, from rabbit