Merck
  • 1,8-Cineole ameliorates oxygen-glucose deprivation/reoxygenation-induced ischaemic injury by reducing oxidative stress in rat cortical neuron/glia.
1,8-Cineole ameliorates oxygen-glucose deprivation/reoxygenation-induced ischaemic injury by reducing oxidative stress in rat cortical neuron/glia.
The Journal of pharmacy and pharmacology (2014-08-05)
Sangwoo Ryu, Hyeon Park, Geun Hee Seol, In-Young Choi
ABSTRACT
1,8-Cineole, the main monoterpene in many essential oils, has been used as an ingredient in flavourings and medicine. 1,8-Cineole has been shown to possess pharmacological properties, including anti-oxidative, anti-inflammatory and anti-nociceptive actions. However, to date, no studies have examined the potential of 1,8-cineole to protect against cerebral ischaemic injury. In this study, we investigated the neuroprotective effects of 1,8-cineole against cortical neuronal/glial cell injury caused by oxygen-glucose deprivation/reoxygenation (OGD/R) in an in-vitro model of ischaemia. 1,8-Cineole significantly attenuated OGD/R-induced cortical cell injury, as well as reduced n-methyl-d-aspartate (NMDA)-induced cell injury. However, it did not inhibit NMDA-induced cytosolic calcium overload. Nevertheless, 1,8-cineole significantly reduced the OGD/R- and NMDA-induced overproduction of reactive oxygen species (ROS). These results indicate that 1,8-cineole exerts neuroprotection through its anti-oxidative rather than its anti-excitotoxic, properties. The decrease in OGD/R-induced intracellular superoxide in 1,8-cineole-treated cortical cells was associated with the upregulation of superoxide dismutase activity. Moreover, 1,8-cineole showed direct ROS scavenging activity in an assay of oxygen radical absorbance capacity. Collectively, these results suggest 1,8-cineole as a potentially effective neuroprotective and anti-oxidative candidate for the treatment of patients with ischaemic stroke.
Product Description
Supelco
1,8-Cineole, primary reference standard
Sigma-Aldrich
Formaldehyde-12C solution, 20% in H2O, 99.9 atom % 12C
Sigma-Aldrich
Eucalyptol, tested according to Ph. Eur.
SAFC
Formaldehyde solution, contains 10-15% methanol as stabilizer, 37 wt. % in H2O
Cineole, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Formaldehyde solution, tested according to Ph. Eur.
Supelco
Eucalyptol, analytical standard
Sigma-Aldrich
Dihydroethidium, BioReagent, suitable for fluorescence, ≥95% (HPCE)
Sigma-Aldrich
Formaldehyde solution, meets analytical specification of USP, ≥34.5 wt. %
Sigma-Aldrich
Formaldehyde solution, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
Sigma-Aldrich
Eucalyptol, natural, ≥99%, FCC, FG
Sigma-Aldrich
Formaldehyde solution, Molecular Biology, BioReagent, ≥36.0% in H2O (T)
Supelco
Formaldehyde solution, stabilized with methanol, ~37 wt. % in H2O, certified reference material
Sigma-Aldrich
Dihydroethidium, ≥95%
Sigma-Aldrich
Formaldehyde solution, Molecular Biology, 36.5-38% in H2O
Sigma-Aldrich
Eucalyptol, 99%