Merck
  • An in vitro metabolic system of gut flora and the metabolism of ginsenoside Rg3 and cholic acid.
An in vitro metabolic system of gut flora and the metabolism of ginsenoside Rg3 and cholic acid.
European journal of drug metabolism and pharmacokinetics (2013-06-12)
Chunyan Zhao, Runbin Sun, Bei Cao, Shenghua Gu, Jieyu Zhao, Linsheng Liu, Xinwen Wang, Weibin Zha, Xiaoyi Yu, Wenjing Xiao, Yong Mao, Chun Ge, Jiaqi Ju, Lixiang Aa, Fei Fei, Yi Ding, Jiye Aa, Guangji Wang
ABSTRACT
For orally administered drugs, the metabolism of a drug by the gut flora plays an important role in the bioavailability, activation and disposition of the drug in vivo. However, no in vitro system is currently available to evaluate the metabolism of a drug by the gut flora before the drug is absorbed into the body. This paper presents an in vitro metabolic system in an anaerobic environment that could be used to evaluate the metabolism of an endogenous compound, cholic acid, and a xenobiotic compound, ginsenoside Rg3. We showed that the proliferation of the anaerobic bacteria of the gut content of hamsters produced a similar composition of gut flora in a culture medium for yeast to that in vivo. Incubation of ginsenoside Rg3 and cholic acid in the anaerobic in vitro system efficiently produced the metabolites Rh2 and deoxycholic acid, respectively, similar to those seen in the gut content in vivo. In comparison with in vivo analysis, this anaerobic in vitro metabolic system is convenient, reproducible, economic and animal saving, and can easily be applied to assess the transformation and disposition of a drug before it enters into the circulatory system.
Sigma-Aldrich
L-Cysteine, Wacker Chemie AG, ≥98.0%
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
USP
Methyl alcohol, United States Pharmacopeia (USP) Reference Standard
USP
Residual Solvent Class 2 - Acetonitrile, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
L-Cysteine, 97%
Sigma-Aldrich
Sodium hydroxide-16O solution, 20 wt. % in H216O, 99.9 atom % 16O
Sigma-Aldrich
L-Cysteine, from non-animal source, BioReagent, suitable for cell culture, ≥98%
Sigma-Aldrich
L-Cysteine, ≥97%, FG
Sigma-Aldrich
L-Cysteine, BioUltra, ≥98.5% (RT)
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
SAFC
L-Cysteine
Sigma-Aldrich
Methanol solution, suitable for NMR (reference standard), 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Supelco
Acetonitrile, analytical standard
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
Supelco
L-Cysteine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Diethyl ether, contains 1 ppm BHT as inhibitor, anhydrous, ≥99.7%
Sigma-Aldrich
Heptane, anhydrous, 99%
Sigma-Aldrich
Heptane, biotech. grade, ≥99%
Sigma-Aldrich
Diethyl ether
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Supelco
Sodium hydroxide solution, 49-51% in water, eluent for IC
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, Molecular Biology, 10 M in H2O
Sigma-Aldrich
Sodium hydroxide, BioUltra, suitable for luminescence, ≥98.0% (T), pellets