Merck
  • Apoptotic effects of platelet factor VIII on megakaryopoiesis: implications for a modified human FVIII for platelet-based gene therapy.
Apoptotic effects of platelet factor VIII on megakaryopoiesis: implications for a modified human FVIII for platelet-based gene therapy.
Journal of thrombosis and haemostasis : JTH (2014-10-08)
T K Greene, R B Lyde, S C Bailey, M P Lambert, L Zhai, D E Sabatino, R M Camire, V R Arruda, M Poncz
ABSTRACT
Ectopically expressed B-domainless factor VIII in megakaryocytes is stored in α-granules, is effective in a number of murine hemostatic models, and is protected from circulating inhibitors. However, this platelet (p) FVIII has different temporal-spatial availability from plasma FVIII, with limited efficacy in other murine hemostatic models. We sought to improve pFVIII hemostatic efficacy by expressing canine (c) FVIII, which has higher stability and activity than human (h) FVIII in FVIII(null) mice. We found that pcFVIII was more effective than phFVIII at restoring hemostasis, but peak pcFVIII antigen levels were lower and were associated with greater megakaryocyte apoptosis than phFVIII. These new insights suggest that pFVIII gene therapy strategies should focus on enhancing activity rather than levels. We previously showed that modification of the PACE/furin cleavage site in hFVIII resulted in secretion of hFVIII primarily as a single-chain molecule with increased biological activity. In megakaryocytes, this variant was expressed at the same level as phFVIII with a lentiviral bone marrow transplant approach to reconstitute FVIII(null) mice, but was more effective, resulting in near-normal hemostasis in the cremaster laser injury model. These studies may have implications for pFVIII gene therapy in hemophilia A.
Product Description
Supelco
Theophylline, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Citrate Concentrated Solution, BioUltra, Molecular Biology, 1 M in H2O
Sigma-Aldrich
Citrate Concentrated Solution, BioReagent, suitable for coagulation assays, 4 % (w/v)
Sigma-Aldrich
2-Mercaptoethanol, ≥99.0%
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, suitable for protein labeling, ≥90% (HPLC), powder
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, ≥97.5% (HPLC)
USP
Theophylline, United States Pharmacopeia (USP) Reference Standard
Theophylline, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
3-tert-Butyl-4-hydroxyanisole, ≥98% (sum of isomers, GC), ≤10% 2-BHA basis (GC)
Sigma-Aldrich
2-Mercaptoethanol, Molecular Biology, suitable for electrophoresis, suitable for cell culture, BioReagent, 99% (GC/titration)
Sigma-Aldrich
Theophylline, anhydrous, ≥99%, powder
Sigma-Aldrich
2-Mercaptoethanol, BioUltra, Molecular Biology, ≥99.0% (GC)
Supelco
2-Mercaptoethanol, derivatization grade (HPLC), LiChropur, ≥99.0% (GC)
Supelco
Sodium Citrate, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
3-tert-Butyl-4-hydroxyanisole, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Adenosine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium butyrate, ≥98.5% (GC)
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, ≥90% (HPLC)
Sigma-Aldrich
Sodium butyrate, 98%
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, BioReagent, suitable for fluorescence, mixture of 2 components, ≥90% (HPLC)
Adenosine, European Pharmacopoeia (EP) Reference Standard
Supelco
Sodium butyrate, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Adenosine
Sigma-Aldrich
Adenosine, BioReagent, suitable for cell culture
Sigma-Aldrich
Adenosine, ≥99%