Merck
  • Localized adenosine signaling provides fine-tuned negative feedback over a wide dynamic range of neocortical network activities.
Localized adenosine signaling provides fine-tuned negative feedback over a wide dynamic range of neocortical network activities.
Journal of neurophysiology (2014-11-14)
Mark J Wall, Magnus J E Richardson
ABSTRACT
Although the patterns of activity produced by neocortical networks are now better understood, how these states are activated, sustained, and terminated still remains unclear. Negative feedback by the endogenous neuromodulator adenosine may potentially play an important role, as it can be released by activity and there is dense A1 receptor expression in the neocortex. Using electrophysiology, biosensors, and modeling, we have investigated the properties of adenosine signaling during physiological and pathological network activity in rat neocortical slices. Both low- and high-rate network activities were reduced by A1 receptor activation and enhanced by block of A1 receptors, consistent with activity-dependent adenosine release. Since the A1 receptors were neither saturated nor completely unoccupied during either low- or high-rate activity, adenosine signaling provides a negative-feedback mechanism with a wide dynamic range. Modeling and biosensor experiments show that during high-rate activity increases in extracellular adenosine concentration are highly localized and are uncorrelated over short distances that are certainly<500 μm. Modeling also predicts that the slow rise of the purine waveform cannot be from diffusion from distal release sites but more likely results from uptake and metabolism. The inability to directly measure adenosine release during low-rate activity, although it is present, is probably a consequence of small localized increases in adenosine concentration that are rapidly diminished by diffusion and active removal mechanisms. Saturation of such removal mechanisms when higher concentrations of adenosine are released results in the accumulation of inosine, explaining the strong purine signal during high-rate activity.
Sigma-Aldrich
HEPES buffer solution, 1 M in H2O
Sigma-Aldrich
Adenosine
Sigma-Aldrich
Inosine, ≥99% (HPLC)
Sigma-Aldrich
Potassium gluconate, 97.0-103.0% dry basis, meets USP testing specifications
Sigma-Aldrich
Xanthine, BioUltra, ≥99%
Sigma-Aldrich
Adenosine, ≥99%
Sigma-Aldrich
Xanthine, ≥99%
Sigma-Aldrich
Potassium D-gluconate, ≥99% (HPLC)
Sigma-Aldrich
Adenosine, BioReagent, suitable for cell culture
Adenosine, European Pharmacopoeia (EP) Reference Standard
Supelco
Adenosine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Potassium gluconate, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Potassium gluconate, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Xanthine, ≥99.5% (HPLC), purified by recrystallization
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Sigma-Aldrich
HEPES, BioUltra, Molecular Biology, ≥99.5% (T)
Sigma-Aldrich
Sodium chloride solution, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Hypoxanthine, ≥99.0%
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Biocytin, ≥98% (TLC)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)