Merck
  • Absolute quantification of superoxide dismutase in cytosol and mitochondria of mice hepatic cells exposed to mercury by a novel metallomic approach.
Absolute quantification of superoxide dismutase in cytosol and mitochondria of mice hepatic cells exposed to mercury by a novel metallomic approach.
Analytica chimica acta (2014-08-17)
M A García-Sevillano, T García-Barrera, F Navarro, J L Gómez-Ariza
ABSTRACT
In the last years, the development of new methods for analyzing accurate and precise individual metalloproteins is of increasing importance, since numerous metalloproteins are excellent biomarkers of oxidative stress and diseases. In that way, methods based on the use of post column isotopic dilution analysis (IDA) or enriched protein standards are required to obtain a sufficient degree of accuracy, precision and high limits of detection. This paper reports the identification and absolute quantification of Cu,Zn-superoxide dismutase (Cu,Zn-SOD) in cytosol and mitochondria from mice hepatic cells using a innovative column switching analytical approach. The method consisted of orthogonal chromatographic systems coupled to inductively coupling plasma-mass spectrometry equipped with a octopole reaction systems (ICP-ORS-MS) and UV detectors: size exclusion fractionation (SEC) of the cytosolic and mitochondrial extracts followed by online anion exchange chromatographic (AEC) separation of Cu/Zn containing species. After purification, Cu,Zn-SOD was identified after tryptic digestion by molecular mass spectrometry (MS). The MS/MS spectrum of a doubly charged peptide was used to obtain the sequence of the protein using the MASCOT searching engine. This optimized methodology reduces the time of analysis and avoids the use of sample preconcentration and clean-up procedures, such as cut-off centrifuged filters, solid phase extraction (SPE), precipitation procedures, off-line fractions insolates, etc. In this sense, the method is robust, reliable and fast with typical chromatographic run time less than 20 min. Precision in terms of relative standard deviation (n = 5) is of 3-5% and detection limits is 0.21 ngCug(-1). The application of the methodology to hepatic cells from mice exposed to inorganic mercury reveals decreased levels of Cu,Zn-SOD in cytosolic and mitochondrial extracts, as a consequence of the oxidative stress caused by this toxic metal. Additionally, the quantification of mitochondrial Cu,Zn-SOD in hepatic cells from Mus musculus has been carried out for the first time.
Sigma-Aldrich
Tris(2-carboxyethyl)phosphine hydrochloride, BioUltra, suitable for electrophoresis, SDS-PAGE tested
Sigma-Aldrich
Tris(2-carboxyethyl)phosphine hydrochloride, powder
Sigma-Aldrich
Tris(2-carboxyethyl)phosphine hydrochloride, BioUltra, ≥98% (NMR)
Sigma-Aldrich
Urea solution, 40 % (w/v) in H2O
Sigma-Aldrich
Urea-12C, 99.9 atom % 12C
Millipore
Urea solution, suitable for microbiology, 40% in H2O
Sigma-Aldrich
Urea solution, BioUltra, ~8 M in H2O
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥98.5% (GC)
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥99.0% (T)
SAFC
Iodoacetamide
Sigma-Aldrich
Iodoacetamide, Single use vial of 56 mg
Urea, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Iodoacetamide, BioUltra
Sigma-Aldrich
Iodoacetamide, ≥99% (NMR), crystalline
Supelco
Urea, 8 M (after reconstitution with 16 mL high purity water)
Sigma-Aldrich
Urea, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99%
Sigma-Aldrich
Urea, ACS reagent, 99.0-100.5%
Sigma-Aldrich
Urea, BioUltra, Molecular Biology, 99% (T)
Sigma-Aldrich
Urea, meets USP testing specifications
Sigma-Aldrich
Urea, BioXtra, pH 7.5-9.5 (20 °C, 5 M in H2O)
Sigma-Aldrich
Urea, ReagentPlus®, ≥99.5%, pellets
Sigma-Aldrich
Urea, suitable for electrophoresis
Supelco
Urea, analytical standard
Sigma-Aldrich
Urea, powder, BioReagent, Molecular Biology, suitable for cell culture
USP
Urea, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Urea, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%, 99.0-101.0% (calc. on dry substance)
Supelco
Tris(2-carboxyethyl)phosphine hydrochloride solution, 0.5 M, pH 7.0(aqueous solution; pH was adjusted with ammonium hydroxide)