Merck
  • Aurora kinase B is important for antiestrogen resistant cell growth and a potential biomarker for tamoxifen resistant breast cancer.
Aurora kinase B is important for antiestrogen resistant cell growth and a potential biomarker for tamoxifen resistant breast cancer.
BMC cancer (2015-04-18)
Sarah L Larsen, Christina W Yde, Anne-Vibeke Laenkholm, Birgitte B Rasmussen, Anne Katrine Duun-Henriksen, Martin Bak, Anne E Lykkesfeldt, Tove Kirkegaard
ABSTRACT
Resistance to antiestrogen therapy is a major clinical challenge in the treatment of estrogen receptor α (ER)-positive breast cancer. The aim of the study was to explore the growth promoting pathways of antiestrogen resistant breast cancer cells to identify biomarkers and novel treatment targets. Antiestrogen sensitive and resistant T47D breast cancer cell lines were used as model systems. Parental and fulvestrant resistant cell lines were subjected to a kinase inhibitor library. Kinase inhibitors preferentially targeting growth of fulvestrant resistant cells were identified and the growth inhibitory effect verified by dose-response cell growth experiments. Protein expression and phosphorylation were investigated by western blot analysis. Cell cycle phase distribution and cell death were analyzed by flow cytometry. To evaluate Aurora kinase B as a biomarker for endocrine resistance, immunohistochemistry was performed on archival primary tumor tissue from breast cancer patients who have received adjuvant endocrine treatment with tamoxifen. The selective Aurora kinase B inhibitor barasertib was identified to preferentially inhibit growth of fulvestrant resistant T47D breast cancer cell lines. Compared with parental cells, phosphorylation of Aurora kinase B was higher in the fulvestrant resistant T47D cells. Barasertib induced degradation of Aurora kinase B, caused mitotic errors, and induced apoptotic cell death as measured by accumulation of SubG1 cells and PARP cleavage in the fulvestrant resistant cells. Barasertib also exerted preferential growth inhibition of tamoxifen resistant T47D cell lines. Finally, high percentage of Aurora kinase B positive tumor cells was significantly associated with reduced disease-free and overall survival in 261 ER-positive breast cancer patients, who have received tamoxifen as first-line adjuvant endocrine treatment. Our results indicate that Aurora kinase B is a driving factor for growth of antiestrogen resistant T47D breast cancer cell lines, and a biomarker for reduced benefit of tamoxifen treatment. Thus, inhibition of Aurora kinase B, e.g. with the highly selective kinase inhibitor barasertib, could be a candidate new treatment for breast cancer patients with acquired resistance to antiestrogens.
Sigma-Aldrich
Fulvestrant, >98% (HPLC)
Supelco
Hydrogen peroxide solution, 30 % (w/w), for ultratrace analysis
Sigma-Aldrich
Formaldehyde-12C solution, 20% in H2O, 99.9 atom % 12C
Sigma-Aldrich
Hydrogen peroxide solution, 30 % (w/w) in H2O, contains stabilizer
Millipore
Hydrogen peroxide solution, 3%, suitable for microbiology
Sigma-Aldrich
Formaldehyde solution, Molecular Biology, BioReagent, ≥36.0% in H2O (T)
Sigma-Aldrich
Formaldehyde solution, meets analytical specification of USP, ≥34.5 wt. %
Supelco
Hydrogen peroxide solution, ≥30%, for trace analysis
Sigma-Aldrich
Hydrogen peroxide solution, 34.5-36.5%
Sigma-Aldrich
Hydrogen peroxide solution, tested according to Ph. Eur.
Sigma-Aldrich
Formaldehyde solution, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
Sigma-Aldrich
Propidium iodide, ≥94% (HPLC)
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Sigma-Aldrich
Formaldehyde solution, tested according to Ph. Eur.
Sigma-Aldrich
Formaldehyde solution, Molecular Biology, 36.5-38% in H2O
Sigma-Aldrich
Monoclonal Anti-AURKB antibody produced in mouse, clone 13E8D3, ascites fluid
SAFC
Formaldehyde solution, contains 10-15% methanol as stabilizer, 37 wt. % in H2O
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 30 wt. % in H2O, meets USP testing specifications
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 30 wt. % in H2O, ACS reagent
Sigma-Aldrich
Ethyl alcohol, Pure 190 proof, for molecular biology
Sigma-Aldrich
Hydrogen peroxide solution, 50 wt. % in H2O, stabilized
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
USP
Dehydrated Alcohol, United States Pharmacopeia (USP) Reference Standard
Supelco
4-tert-Octylphenol monoethoxylate solution, 10 μg/mL in acetone, analytical standard
Supelco
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Formaldehyde solution, stabilized with methanol, ~37 wt. % in H2O, certified reference material
Fulvestrant, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Ethanol, BioUltra, Molecular Biology, ≥99.8%, (absolute alcohol, without additive, A15 o1)
Sigma-Aldrich
Propidium iodide solution
Sigma-Aldrich
Ethanol, tested according to Ph. Eur.