Merck
  • Combined treatment with ABT-737 and VX-680 induces apoptosis in Bcl-2- and c-FLIP-overexpressing breast carcinoma cells.
Combined treatment with ABT-737 and VX-680 induces apoptosis in Bcl-2- and c-FLIP-overexpressing breast carcinoma cells.
Oncology reports (2015-01-17)
Jung Eun Choi, Seon Min Woo, Kyoung-Jin Min, Su Hwan Kang, Soo Jung Lee, Taeg Kyu Kwon
ABSTRACT
ABT-737, a BH3-mimetic small-molecule inhibitor, binds with very high affinity to Bcl-2, Bcl-xL and Bcl-w, and inhibits their activity. Aurora kinase is one of the serine/threonine kinase family members and is a vital and critical regulator of mitosis and meiosis. In the present study, we investigated the effects and mechanisms of a combined treatment of ABT-737 and VX-680 (Aurora kinase inhibitor) in human breast cancer MDA-MB‑435S cells. ABT-737 plus VX-680 induced caspase-dependent apoptosis in the human breast cancer cells. Combined treatment with ABT-737 and VX-680 led to the downregulation of Bcl-2 expression at the transcriptional level and the downregulation of c-FLIP and Mcl-1 expression at the post-transcriptional level. Overexpression of Bcl-2 or c-FLIP could not block the induction of apoptosis caused by the combined treatment with ABT-737 and VX-680. However, overexpression of Mcl-1 partially inhibited the induction of apoptosis. In contrast, the combined treatment with ABT-737 and VX680 had no effect on the apoptosis in normal cells. Taken together, our study demonstrated that combined treatment with ABT-737 and VX-680 induced apoptosis in anti‑apoptotic protein (Bcl-2 or c-FLIP)-overexpressing cells.
Sigma-Aldrich
Glycerol solution, 83.5-89.5% (T)
USP
Glycerin, United States Pharmacopeia (USP) Reference Standard
Supelco
Glycerin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Glycerol, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
Sigma-Aldrich
Glycerol, BioXtra, ≥99% (GC)
Sigma-Aldrich
Glycerol, Molecular Biology, ≥99.0%
Sigma-Aldrich
Adenosine 3′,5′-cyclic monophosphate tris salt, ≥97% (HPLC), powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Glycerol, ≥99.5%
Sigma-Aldrich
Glycerin, meets USP testing specifications
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, ≥99.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, BioUltra, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Glycerol, tested according to Ph. Eur., anhydrous
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Glycerol, ReagentPlus®, ≥99.0% (GC)
Sigma-Aldrich
Adenosine 3′,5′-cyclic monophosphate, ≥98.5% (HPLC), powder
Sigma-Aldrich
Glycerol, BioUltra, Molecular Biology, anhydrous, ≥99.5% (GC)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Glycerol, FCC, FG
Sigma-Aldrich
Glycerol, ACS reagent, ≥99.5%
Supelco
Glycerol, analytical standard