Merck
  • Targeting poly(ADP-ribose) polymerase and the c-Myb-regulated DNA damage response pathway in castration-resistant prostate cancer.
Targeting poly(ADP-ribose) polymerase and the c-Myb-regulated DNA damage response pathway in castration-resistant prostate cancer.
Science signaling (2014-05-23)
Likun Li, Wenjun Chang, Guang Yang, Chengzhen Ren, Sanghee Park, Theodoros Karantanos, Styliani Karanika, Jianxiang Wang, Jianhua Yin, Parantu K Shah, Hirayama Takahiro, Masato Dobashi, Wenling Zhang, Eleni Efstathiou, Sankar N Maity, Ana M Aparicio, Elsa M Li Ning Tapia, Patricia Troncoso, Bradley Broom, Lianchun Xiao, Hyun-Sung Lee, Ju-Seog Lee, Paul G Corn, Nora Navone, Timothy C Thompson
ABSTRACT
Androgen deprivation is the standard treatment for advanced prostate cancer (PCa), but most patients ultimately develop resistance and tumor recurrence. We found that MYB is transcriptionally activated by androgen deprivation therapy or genetic silencing of the androgen receptor (AR). MYB silencing inhibited PCa growth in culture and xenografts in mice. Microarray data revealed that c-Myb and AR shared a subset of target genes that encode DNA damage response (DDR) proteins, suggesting that c-Myb may supplant AR as the dominant regulator of their common DDR target genes in AR inhibition-resistant or AR-negative PCa. Gene signatures including AR, MYB, and their common DDR-associated target genes positively correlated with metastasis, castration resistance, tumor recurrence, and decreased survival in PCa patients. In culture and in xenograft-bearing mice, a combination strategy involving the knockdown of MYB, BRCA1, or TOPBP1 or the abrogation of cell cycle checkpoint arrest with AZD7762, an inhibitor of the checkpoint kinase Chk1, increased the cytotoxicity of the poly[adenosine 5'-diphosphate (ADP)-ribose] polymerase (PARP) inhibitor olaparib in PCa cells. Our results reveal new mechanism-based therapeutic approaches for PCa by targeting PARP and the DDR pathway involving c-Myb, TopBP1, ataxia telangiectasia mutated- and Rad3-related (ATR), and Chk1.
Sigma-Aldrich
DL-Glyceraldehyde 3-phosphate solution, 45-55 mg/mL in H2O
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Myb
Sigma-Aldrich
ApopTag Plus Peroxidase In Situ Apoptosis Kit, The ApopTag Plus Peroxidase In Situ Apoptosis Detection Kit detects apoptotic cells by labeling & detecting DNA strand breaks by the indirect TUNEL method.
Sigma-Aldrich
Chromatin Immunoprecipitation (ChIP) Assay Kit, Contains all necessary reagents to perform 22 individual chromatin immunoprecipitation (ChIP) reactions using inexpensive protein A agarose beads.
Sigma-Aldrich
Anti-acetyl-Histone H3 (Lys27) Antibody, serum, Upstate®
Sigma-Aldrich
MISSION® esiRNA, targeting human MYB
Sigma-Aldrich
Anti-trimethyl-Histone H3 (Lys27) Antibody, Upstate®, from rabbit