Merck
  • Prior high corticosterone exposure reduces activation of immature neurons in the ventral hippocampus in response to spatial and nonspatial memory.
Prior high corticosterone exposure reduces activation of immature neurons in the ventral hippocampus in response to spatial and nonspatial memory.
Hippocampus (2014-10-02)
Joanna L Workman, Melissa Y T Chan, Liisa A M Galea
ABSTRACT
Chronic stress or chronically high glucocorticoids attenuate adult hippocampal neurogenesis by reducing cell proliferation, survival, and differentiation in male rodents. Neurons are still produced in the dentate gyrus during chronically high glucocorticoids, but it is not known whether these new neurons are appropriately activated in response to spatial memory. Thus, the goal of this study was to determine whether immature granule neurons generated during chronically high glucocorticoids (resulting in a depressive-like phenotype) are differentially activated by spatial memory retrieval. Male Sprague Dawley rats received either 40 mg/kg corticosterone (CORT) or vehicle for 18 days prior to behavioral testing. Rats were tested in the forced swim test (FST) and then tested in a spatial (hippocampus-dependent) or cued (hippocampus-independent) Morris Water Maze. Tissue was then processed for doublecortin (DCX) to identify immature neurons and zif268, an immediate early gene product. As expected, CORT increased depressive-like behavior (greater immobility in the FST) however, prior CORT modestly enhanced spatial learning and memory compared with oil. Prior CORT reduced the number of DCX-expressing cells and proportion of DCX-expressing cells colabeled for zif268, but only in the ventral hippocampus. Prior CORT shifted the proportion of cells in the ventral hippocampus away from postmitotic cells and toward immature, proliferative cells, likely due to the fact that postmitotic cells were produced and matured during CORT exposure but proliferative cells were produced after high CORT exposure ceased. Compared with cue training, spatial training slightly increased DCX-expressing cells and shifted cells toward the postmitotic stage in the ventral hippocampus. These data suggest that the effects of CORT and spatial training on immature neurons are more pronounced in the ventral hippocampus. Further, high CORT reduced activation of immature neurons, suggesting that exposure to high CORT may have long-term effects on cell integration or function.
Product Description
USP
Glycerin, United States Pharmacopeia (USP) Reference Standard
Supelco
Glycerin, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Ethylene glycol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Tris(tert-butoxy)silanol, 99.999%
Sigma-Aldrich
Glycerol solution, 83.5-89.5% (T)
Sigma-Aldrich
Ethylene glycol, BioUltra, ≥99.5% (GC)
Supelco
Ethylene glycol, analytical standard
Sigma-Aldrich
Ethylene glycol, anhydrous, 99.8%
USP
Ethylene glycol, United States Pharmacopeia (USP) Reference Standard
Supelco
Ethylene glycol solution, suitable for NMR (reference standard), 80% in DMSO-d6 (99.9 atom % D), NMR tube size 5 mm × 8 in.
Sigma-Aldrich
Ethylene glycol, ReagentPlus®, ≥99%
Sigma-Aldrich
Ethylene glycol, spectrophotometric grade, ≥99%
Sigma-Aldrich
Glycerol, FCC, FG
USP
Sucrose, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Sucrose, puriss., meets analytical specification of Ph. Eur., BP, NF
Millipore
Sucrose, ACS reagent, suitable for microbiology, ≥99.0%
Sigma-Aldrich
Glycerol, BioUltra, Molecular Biology, anhydrous, ≥99.5% (GC)
Sigma-Aldrich
Glycerol, tested according to Ph. Eur., anhydrous
Sigma-Aldrich
Sucrose, BioUltra, Molecular Biology, ≥99.5% (HPLC)
Sigma-Aldrich
Glycerol, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
Sigma-Aldrich
Glycerol, Molecular Biology, ≥99.0%
Sigma-Aldrich
Glycerin, meets USP testing specifications
Sigma-Aldrich
Glycerol, BioXtra, ≥99% (GC)
Sigma-Aldrich
Glycerol, ≥99.5%
Sigma-Aldrich
Sucrose, ACS reagent
Sigma-Aldrich
Sucrose, ≥99.5% (GC), BioXtra
Sigma-Aldrich
Sucrose, meets USP testing specifications
Supelco
Sucrose, analytical standard, for enzymatic assay kit SCA20
Sigma-Aldrich
Sucrose, ≥99% (GC), Grade I, suitable for plant cell culture
Sigma-Aldrich
Sucrose, ≥99.5% (GC)